Search
Search

Esrp1| Gene Symbol: Esrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290383.1 | NP_001277312.1 | 9 - 10 | 1281 | 67 | |
| NM_194055.3 | NP_918944.2 | 9 - 10 | 1284 | 67 | ||
| GenBank mRNA | AK042164.1 | - | 9 - 10 | 896 | 67 | |
| AK140848.1 | - | 9 - 10 | 1282 | 67 | ||
| BC031468.1 | - | 7 - 8 | 587 | 4037840 | 67 | |
| BC059280.1 | - | 7 - 8 | 585 | 67 | ||
| DV654302.1 | - | 4 - 5 | 345 | 67 | ||
| DV659240.1 | - | 7 - 8 | 640 | 67 | ||
Set Membership: |
|