Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839542_g1
          See other SOX15 GE Assays ›
          Gene Symbol:
          Sox15
          Gene Name:
          SRY (sex determining region Y)-box 15
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 69655037 - 69656727 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839542_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sox15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009235.2 NP_033261.1 1 - 2 856 84
          GenBank mRNA AF182945.1 - 1 - 2 520 84
          AK131957.1 - 1 - 2 858 84
          BC119067.1 - 1 - 2 580 8733816 84
          BC119069.1 - 1 - 2 580 8733818 84
          X98369.1 - 1 - 2 247 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000919.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sox15

          Panther Classification:

          Molecular Function -

          HMG box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of satellite cell activation involved in skeletal muscle regeneration
          cell differentiation
          skeletal muscle tissue regeneration
          negative regulation of striated muscle tissue development
          positive regulation of transcription from RNA polymerase II promoter
          myoblast development
          positive regulation of G0 to G1 transition
          positive regulation of myoblast proliferation
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          chromatin binding
          protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896329 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094691 BC119067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094691 BC119069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094691 AK131957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094691 X98369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094691 NM_009235.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164408 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094691 BC119067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094691 BC119069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094691 AK131957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094691 X98369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094691 NM_009235.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440440 BC119067.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440440 BC119069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440440 AK131957.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440440 X98369.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440440 NM_009235.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896309 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164408 - -
          Whole Mouse Genome G4122A A_51_P440440 BC119067.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440440 BC119069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440440 AK131957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440440 X98369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440440 NM_009235.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108954 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09229987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09229976 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09229986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09229989 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09229993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09229988 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09230007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09229977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09229988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09230000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896337 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377591 - -
          Affymetrix HT MG-430 PM Array Plate 1452017_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377588 - -
          Mouse Genome 430 2.0 Array 1452017_at - -
          Mouse Genome 430 2.0 Array 1452017_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20670_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20670_at - -
          Affymetrix Mouse Clariom S Array PSR1100007816.mm.1 NM_009235.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007816.mm.1 NM_009235.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007816.mm.1 TC1100000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007817.mm.1 NM_009235.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007817.mm.1 NM_009235.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007817.mm.1 TC1100000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007813.mm.1 NM_009235.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007813.mm.1 NM_009235.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007813.mm.1 TC1100000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007815.mm.1 NM_009235.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007815.mm.1 NM_009235.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007815.mm.1 TC1100000919.mm.1 MTA TC
          Mouse Expression Set 430 1452017_at - -
          Mouse Expression Set 430 1452017_PM_at - -
          Murine Genome U74v2 Set 96824_PM_at - -
          Murine Genome U74v2 Set 96824_at - -
          Mouse Genome 430A 2.0 Array 1452017_at - -
          Mouse Genome 430A 2.0 Array 1452017_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452017_at - -
          Affymetrix HT Mouse Genome 430A Array 1452017_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 818349 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470603 - -
          Illumina mouse-6 v1.0 expression beadchip 2470603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662811 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2662811 - -
          mouse-6 v1.1 (Illumina) scl020670.2_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020670.2_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662811 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662811 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline