Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839568_m1
          See other SPT1 GE Assays ›
          Gene Symbol:
          Spt1
          Gene Name:
          salivary protein 1
          Gene Aliases:
          Spt-1
          Chromosome Location:
          Chr.15: 103895855 - 103899312 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839568_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009267.2 NP_033293.2 3 - 4 438 120
          GenBank mRNA AK006124.1 - 3 - 4 436 120
          BC030164.1 - 3 - 4 120
          BC106151.1 - 3 - 4 422 4923478 120
          CK616337.1 - 1 - 2 120
          CK616968.1 - 3 - 4 439 120
          M33974.1 - 3 - 4 120
          X00349.1 - 3 - 4 120

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spt1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 BC106151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 CK616337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 CK616968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 BC030164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 M33974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 BC106151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 CK616337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 CK616968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 BC030164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P245277 M33974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 BC106151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 CK616337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 CK616968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 BC030164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 M33974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 BC106151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 CK616337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 CK616968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 BC030164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P245277 M33974.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09578733 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09578741 - -
          Whole Mouse Genome G4122A A_52_P245277 BC106151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 CK616337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 CK616968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 BC030164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 M33974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 BC106151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 CK616337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 CK616968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 BC030164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P245277 M33974.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09578733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433188 - -
          Affymetrix HT MG-430 PM Array Plate 1448168_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433185 - -
          Mouse Genome 430 2.0 Array 1448168_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448168_a_at - -
          Mouse Expression Set 430 1448168_a_at - -
          Mouse Expression Set 430 1448168_PM_a_at - -
          Murine Genome U74v2 Set 94152_PM_at - -
          Murine Genome U74v2 Set 94152_at - -
          Murine Genome U74v2 Set 162108_f_at - -
          Murine Genome U74v2 Set 162108_PM_f_at - -
          Murine Genome U74v2 Set 94153_PM_g_at - -
          Murine Genome U74v2 Set 94153_g_at - -
          Mouse Genome 430A 2.0 Array 1448168_a_at - -
          Mouse Genome 430A 2.0 Array 1448168_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448168_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448168_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4280095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773740 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773740 - -
          mouse-6 v1.1 (Illumina) scl020770.2_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020770.2_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2845370 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2845370 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2681569 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline