Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839780_m1
          See other KRT27 GE Assays ›
          Gene Symbol:
          Krt27
          Gene Name:
          keratin 27
          Gene Aliases:
          Krt1-c29, mIRSa3.1
          Chromosome Location:
          Chr.11: 99345566 - 99351094 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839780_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010666.1 NP_034796.1 7 - 8 1236 76
          GenBank mRNA AB013607.1 - 7 - 8 1236 76
          AK077384.1 - 7 - 8 1248 76
          BC140996.1 - 7 - 8 1260 9055608 76
          BC140997.1 - 7 - 8 1260 9055609 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003704.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt27

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle morphogenesis
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 BC140997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 BC140996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 NM_010666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 AB013607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 AK077384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 BC140997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 BC140996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 NM_010666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 AB013607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412926 AK077384.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137661 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163974 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 BC140997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 BC140996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 NM_010666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 AB013607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 AK077384.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 BC140997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 BC140996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 NM_010666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 AB013607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412926 AK077384.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 BC140997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 BC140996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 NM_010666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 AB013607.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 AK077384.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 BC140997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 BC140996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 NM_010666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 AB013607.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412926 AK077384.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28061834 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07460942 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163974 - -
          Whole Mouse Genome G4122A A_51_P412926 BC140997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 BC140996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 NM_010666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 AB013607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 AK077384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 BC140997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 BC140996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 NM_010666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 AB013607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P412926 AK077384.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140153 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106452 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163974 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08980029 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09726939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09726940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08980023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08980026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28061816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09238073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09194226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09601914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09194220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28061818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390820 - -
          Affymetrix HT MG-430 PM Array Plate 1449378_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10390816 - -
          Mouse Genome 430 2.0 Array 1449378_at - -
          Mouse Genome 430 2.0 Array 1449378_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16675_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16675_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100034174.mm.1 NM_010666.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034174.mm.1 TC1100003704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034173.mm.1 NM_010666.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034173.mm.1 TC1100003704.mm.1 MTA TC
          Mouse Expression Set 430 1449378_at - -
          Mouse Expression Set 430 1449378_PM_at - -
          Murine Genome U74v2 Set 93800_f_at - -
          Murine Genome U74v2 Set 93800_PM_f_at - -
          Murine Genome U74v2 Set 167556_PM_f_at - -
          Murine Genome U74v2 Set 167973_PM_f_at - -
          Murine Genome U74v2 Set 167556_f_at - -
          Murine Genome U74v2 Set 167973_f_at - -
          Mouse Genome 430A 2.0 Array 1449378_PM_at - -
          Mouse Genome 430A 2.0 Array 1449378_at - -
          Affymetrix HT Mouse Genome 430A Array 1449378_at - -
          Affymetrix HT Mouse Genome 430A Array 1449378_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 796074 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2640333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002640333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613938 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613938 - -
          mouse-6 v1.1 (Illumina) scl39601.8.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 2640333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39601.8.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2640333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613938 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613938 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline