Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839856_g1
          See other SHBG GE Assays ›
          Gene Symbol:
          Shbg
          Gene Name:
          sex hormone binding globulin
          Gene Aliases:
          ABP
          Chromosome Location:
          Chr.11: 69610281 - 69618025 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839856_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shbg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011367.2 NP_035497.1 4 - 5 599 76
          GenBank mRNA AK010959.1 - 4 - 5 599 76
          BC120785.1 - 4 - 5 584 40129708 76
          BC120787.1 - 4 - 5 584 40129710 76
          U85644.1 - 4 - 5 562 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003050.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shbg

          Gene Ontology Categories:

          Function(s) Process(es)

          primary spermatocyte growth
          steroid binding
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 BC120787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 U85644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 NM_011367.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 AK010959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 BC120785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 BC120787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 U85644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 NM_011367.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 AK010959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423549 BC120785.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88831 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148803 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 BC120787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 U85644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 NM_011367.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 AK010959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 BC120785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 BC120787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 U85644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 NM_011367.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 AK010959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423549 BC120785.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896137 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148803 - -
          Whole Mouse Genome G4122A A_51_P423549 BC120787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 U85644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 NM_011367.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 AK010959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 BC120785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 BC120787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 U85644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 NM_011367.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 AK010959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423549 BC120785.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127337 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148803 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08986163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08986167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09007102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09099570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09099571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09271196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08986163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09047740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09271198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09673203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896131 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10387520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387522 - -
          Affymetrix HT MG-430 PM Array Plate 1421602_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10387516 - -
          Mouse Genome 430 2.0 Array 1421602_PM_at - -
          Mouse Genome 430 2.0 Array 1421602_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20415_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20415_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100027884.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027884.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027884.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027892.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027892.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027892.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027893.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027893.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027893.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027886.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027886.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027886.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027883.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027883.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027883.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027891.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027891.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027891.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027882.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027882.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027882.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027887.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027887.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027887.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027888.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027888.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027888.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027885.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027885.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027885.mm.1 TC1100003050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027889.mm.1 NM_011367.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027889.mm.1 NM_011367.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100027889.mm.1 TC1100003050.mm.1 MTA TC
          Mouse Expression Set 430 1421602_PM_at - -
          Mouse Expression Set 430 1421602_at - -
          Murine Genome U74v2 Set 100702_PM_at - -
          Murine Genome U74v2 Set 100702_at - -
          Mouse Genome 430A 2.0 Array 1421602_PM_at - -
          Mouse Genome 430A 2.0 Array 1421602_at - -
          Affymetrix HT Mouse Genome 430A Array 1421602_at - -
          Affymetrix HT Mouse Genome 430A Array 1421602_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 623359 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5340551 - -
          Illumina mouse-6 v1.0 expression beadchip 5340551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227933 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227933 - -
          mouse-6 v1.1 (Illumina) scl40026.8.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40026.8.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227933 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227933 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline