Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839889_m1
          See other STAT5B GE Assays ›
          Gene Symbol:
          Stat5b
          Gene Name:
          signal transducer and activator of transcription 5B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 100780731 - 100859004 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839889_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113563.1 NP_001107035.1 2 - 3 326 92
          NM_011489.3 NP_035619.3 2 - 3 658 92
          GenBank mRNA AK150098.1 - 2 - 3 295 92
          AK154014.1 - 2 - 3 658 92
          AK154664.1 - 2 - 3 292 92
          AY040231.1 - 2 - 3 612 92
          AY042906.1 - 2 - 3 612 92
          AY044901.1 - 2 - 3 612 92
          AY044902.1 - 2 - 3 612 92
          AY044903.1 - 2 - 3 612 92
          BC024319.1 - 2 - 3 282 4237132 92
          U21110.2 - 2 - 3 612 92
          Z48539.1 - 2 - 3 226 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003787.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stat5b

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          allantoin metabolic process
          luteinization
          natural killer cell differentiation
          liver development
          citrate metabolic process
          2-oxoglutarate metabolic process
          succinate metabolic process
          oxaloacetate metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          isoleucine metabolic process
          valine metabolic process
          creatine metabolic process
          fatty acid metabolic process
          acute-phase response
          signal transduction
          JAK-STAT cascade
          sex differentiation
          female pregnancy
          lactation
          positive regulation of cell proliferation
          regulation of steroid metabolic process
          cytokine-mediated signaling pathway
          taurine metabolic process
          lipid storage
          regulation of cell adhesion
          regulation of epithelial cell differentiation
          response to estradiol
          response to lipopolysaccharide
          positive regulation of natural killer cell proliferation
          positive regulation of natural killer cell differentiation
          cellular response to hormone stimulus
          T cell differentiation in thymus
          prolactin signaling pathway
          regulation of multicellular organism growth
          positive regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          progesterone metabolic process
          T cell homeostasis
          negative regulation of apoptotic process
          response to peptide hormone
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of lymphocyte differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of natural killer cell mediated cytotoxicity
          creatinine metabolic process
          development of secondary female sexual characteristics
          development of secondary male sexual characteristics
          Peyer's patch development
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of cellular component movement
          JAK-STAT cascade involved in growth hormone signaling pathway
          response to interleukin-2
          response to interleukin-4
          response to interleukin-15
          cellular response to growth factor stimulus
          cellular response to epidermal growth factor stimulus
          mast cell migration
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          protein binding
          protein phosphatase binding
          glucocorticoid receptor binding
          sequence-specific DNA binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100034932.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034932.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034932.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034932.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034932.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034917.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034917.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034917.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034917.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034917.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034921.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034921.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034921.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034921.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034921.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034931.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034931.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034931.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034931.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034931.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034941.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034941.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034941.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034918.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034918.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034918.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034918.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034918.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034920.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034920.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034920.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034920.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034920.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034926.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034926.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034926.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034926.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034926.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034939.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034939.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034939.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034914.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034914.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034914.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034914.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034914.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034919.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034919.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034919.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034919.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034919.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034923.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034923.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034923.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034923.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034923.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034935.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034935.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034935.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034915.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034915.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034915.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034915.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034915.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034924.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034924.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034924.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034924.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034924.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034927.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034927.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034927.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034927.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034927.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034928.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034928.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034928.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034928.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034928.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034913.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034913.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034913.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034913.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034913.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034916.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034916.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034916.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034916.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034916.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034922.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034922.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034922.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034922.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034922.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034925.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034925.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034925.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034925.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034925.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034930.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034930.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034930.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034930.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034930.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034936.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034936.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034936.mm.1 TC1100003787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034929.mm.1 NM_001113563.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034929.mm.1 NM_001113563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034929.mm.1 NM_011489.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034929.mm.1 NM_011489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034929.mm.1 TC1100003787.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 491384 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline