Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839913_m1
          See other WAP GE Assays ›
          Gene Symbol:
          Wap
          Gene Name:
          whey acidic protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 6635483 - 6638649 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839913_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011709.5 NP_035839.3 1 - 2 111 83
          GenBank mRNA AK142655.1 - 112 83
          AK164735.1 - 1 - 2 112 83
          BC132280.1 - 1 - 2 109 40130557 83
          BC132282.1 - 1 - 2 109 40130559 83
          V00856.1 - 1 - 2 99 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002183.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wap

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          peptidase inhibitor activity
          nutrient reservoir activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407705 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140255 AK142655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140255 AK164735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140255 BC132280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140255 BC132282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140255 V00856.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159605 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110381 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140255 AK142655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140255 AK164735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140255 BC132280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140255 BC132282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140255 V00856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27569541 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07246226 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110381 - -
          Whole Mouse Genome G4122A A_51_P407704 - -
          Whole Mouse Genome G4122A A_51_P407705 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103863 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110381 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09056065 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09056076 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09056080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09056075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09056061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09056070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27569497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09056071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09178670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09026123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09161968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09161971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09583442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09161969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09674980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09026124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10384216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384215 - -
          Affymetrix HT MG-430 PM Array Plate 1448386_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10384212 - -
          Mouse Genome 430 2.0 Array 1448386_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448386_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22373_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22373_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100021292.mm.1 NM_011709.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021292.mm.1 NM_011709.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021292.mm.1 TC1100002183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021293.mm.1 NM_011709.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021293.mm.1 NM_011709.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021293.mm.1 TC1100002183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021298.mm.1 NM_011709.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021298.mm.1 NM_011709.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021298.mm.1 TC1100002183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021290.mm.1 NM_011709.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021290.mm.1 NM_011709.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021290.mm.1 TC1100002183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021295.mm.1 NM_011709.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021295.mm.1 NM_011709.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021295.mm.1 TC1100002183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021291.mm.1 NM_011709.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021291.mm.1 NM_011709.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021291.mm.1 TC1100002183.mm.1 MTA TC
          Mouse Expression Set 430 1448386_a_at - -
          Mouse Expression Set 430 1448386_PM_a_at - -
          Murine Genome U74v2 Set 102931_at - -
          Murine Genome U74v2 Set 102931_PM_at - -
          Mouse Genome 430A 2.0 Array 1448386_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1448386_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448386_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448386_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6020497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232780 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232780 - -
          mouse-6 v1.1 (Illumina) scl40531.6.1_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40531.6.1_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232780 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline