Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840006_m1
          See other PNP GE Assays ›
          Gene Symbol:
          Pnp
          Gene Name:
          purine-nucleoside phosphorylase
          Gene Aliases:
          AL024301, AU015798, Np, Np-1, Np-2, Pnp1
          Chromosome Location:
          Chr.14: 50944303 - 50953412 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840006_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pnp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013632.4 NP_038660.1 2 - 3 458 98
          GenBank mRNA AK049817.1 - 2 - 3 237 98
          AK153451.1 - 2 - 3 460 98
          BC003788.1 - 2 - 3 230 3600212 98
          BC052679.1 - 2 - 3 209 30062908 98
          CT010254.1 - 1 - 2 178 98
          CT010316.1 - 1 - 2 178 98
          L11290.1 - 2 - 3 199 98
          L11291.1 - 2 - 3 199 98
          M84563.1 - 256 98
          U35374.1 - 1 - 2 178 98
          X56548.1 - 2 - 3 255 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pnp

          Panther Classification:

          Molecular Function -

          kinase nucleotide kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          nucleobase-containing compound metabolic process
          inosine catabolic process
          deoxyinosine catabolic process
          deoxyguanosine catabolic process
          GTP biosynthetic process
          intrinsic apoptotic signaling pathway in response to DNA damage
          apoptotic mitochondrial changes
          nucleoside metabolic process
          response to gamma radiation
          urate biosynthetic process
          positive regulation of T cell proliferation
          purine nucleoside metabolic process
          purine-containing compound salvage
          positive regulation of B cell differentiation
          positive regulation of DNA repair
          dGTP metabolic process
          guanosine catabolic process
          positive regulation of alpha-beta T cell differentiation
          negative regulation of T cell apoptotic process
          nucleoside binding
          purine nucleobase binding
          catalytic activity
          purine-nucleoside phosphorylase activity
          drug binding
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring pentosyl groups
          phosphate ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018841 AK049817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018841 NM_013632.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138137 X56548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138137 AK049817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138137 AK153451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138137 NM_013632.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88940 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152643 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018841 AK049817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018841 NM_013632.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138137 X56548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138137 AK049817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138137 AK153451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138137 NM_013632.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488890 X56548.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488890 U35374.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488890 BC052679.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488890 CT010254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488890 CT010316.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488888 X56548.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488888 U35374.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488888 BC052679.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488888 CT010254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488888 CT010316.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152643 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02849317 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08109721 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08109767 - -
          Whole Mouse Genome G4122A A_52_P482090 AK049817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482090 NM_013632.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488888 X56548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488888 U35374.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488888 BC052679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488888 CT010254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488888 CT010316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488890 X56548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488890 U35374.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488890 BC052679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488890 CT010254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488890 CT010316.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138137 X56548.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138137 AK049817.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138137 AK153451.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138137 NM_013632.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11244731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11797620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11797622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11173868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11788367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11625500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599226 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414528 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414527 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414514 - -
          Mouse Genome 430 2.0 Array 1453299_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416530_PM_a_at - -
          Mouse Genome 430 2.0 Array 1453299_a_at - -
          Mouse Genome 430 2.0 Array 1416530_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18950_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18950_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa1220_at - -
          Affymetrix Mouse Clariom S Array PSR1400004497.mm.1 NM_013632.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004497.mm.1 NM_013632.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004497.mm.1 TC1400000574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004502.mm.1 NM_013632.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004502.mm.1 NM_013632.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004502.mm.1 TC1400000574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004496.mm.1 NM_013632.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004496.mm.1 NM_013632.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004496.mm.1 TC1400000574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004498.mm.1 NM_013632.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004498.mm.1 NM_013632.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004498.mm.1 TC1400000574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004500.mm.1 NM_013632.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004500.mm.1 NM_013632.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004500.mm.1 TC1400000574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004499.mm.1 NM_013632.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004499.mm.1 NM_013632.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004499.mm.1 TC1400000574.mm.1 MTA TC
          Mouse Expression Set 430 1416530_a_at - -
          Mouse Expression Set 430 1453299_a_at - -
          Mouse Expression Set 430 1416530_PM_a_at - -
          Mouse Expression Set 430 1453299_PM_a_at - -
          Murine Genome U74v2 Set 93290_at - -
          Murine Genome U74v2 Set 93290_PM_at - -
          Mouse Genome 430A 2.0 Array 1453299_a_at - -
          Mouse Genome 430A 2.0 Array 1416530_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453299_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416530_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 757769 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2630577 - -
          MouseWG-6 v2 Expression BeadChip 3120142 - -
          MouseWG-6 v2 Expression BeadChip 5670068 - -
          MouseWG-6 v2 Expression BeadChip 6290039 - -
          Illumina mouse-6 v1.0 expression beadchip 3120142 - -
          Illumina mouse-6 v1.0 expression beadchip 5670068 - -
          Illumina mouse-6 v1.0 expression beadchip 6290039 - -
          Illumina mouse-6 v1.0 expression beadchip 2630577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290039 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216954 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256633 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215638 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216954 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256633 - -
          mouse-6 v1.1 (Illumina) scl46368.7_1-S - -
          mouse-6 v1.1 (Illumina) scl018950.7_181-S - -
          mouse-6 v1.1 (Illumina) scl0018950.1_0-S - -
          mouse-6 v1.1 (Illumina) scl0018950.1_171-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290039 - -
          Illumina Sentrix mouse-6.v1 bead array 3120142 - -
          Illumina Sentrix mouse-6.v1 bead array 5670068 - -
          Illumina Sentrix mouse-6.v1 bead array 2630577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46368.7_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0018950.1_171-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018950.7_181-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0018950.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670068 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290039 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630577 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256633 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216954 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215638 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256633 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215638 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256020 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216954 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018950.7_181-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline