Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840210_g1
          See other NKX2-4 GE Assays ›
          Gene Symbol:
          Nkx2-4
          Gene Name:
          NK2 homeobox 4
          Gene Aliases:
          1700001P03Rik, Nkx-2.4, tinman
          Chromosome Location:
          Chr.2: 147083337 - 147085439 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840210_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkx2-4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023504.1 NP_075993.1 1 - 2 451 80
          GenBank mRNA AF202039.1 - 1 - 2 248 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004789.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkx2-4

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          multicellular organism development
          DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722283 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722282 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722293 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083103 AF202039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083103 AK005620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083103 NM_023504.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145881 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083103 AF202039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083103 AK005620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083103 NM_023504.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353934 AF202039.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21722338 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145881 - -
          Whole Mouse Genome G4122A A_51_P353934 AF202039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P469165 NM_023504.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118535 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01942358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01942363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01942359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01942364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01752384 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722282 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722283 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21722287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21722329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01867557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01936422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01338538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21722312 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10488363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488364 - -
          Affymetrix HT MG-430 PM Array Plate 1432863_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1432862_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488362 - -
          Mouse Genome 430 2.0 Array 1432863_a_at - -
          Mouse Genome 430 2.0 Array 1432863_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432862_at - -
          Mouse Genome 430 2.0 Array 1432862_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200040961.mm.1 NM_023504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040961.mm.1 NM_023504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040961.mm.1 TC0200004789.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040962.mm.1 NM_023504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040962.mm.1 NM_023504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040962.mm.1 TC0200004789.mm.1 MTA TC
          Mouse Expression Set 430 1432863_a_at - -
          Mouse Expression Set 430 1432862_at - -
          Mouse Expression Set 430 1432863_PM_a_at - -
          Mouse Expression Set 430 1432862_PM_at - -
          Murine Genome U74v2 Set 171243_at - -
          Murine Genome U74v2 Set 171243_PM_at - -
          Mouse Genome 430A 2.0 Array 1432863_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432862_PM_at - -
          Mouse Genome 430A 2.0 Array 1432863_a_at - -
          Mouse Genome 430A 2.0 Array 1432862_at - -
          Affymetrix HT Mouse Genome 430A Array 1432862_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1432863_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432862_at - -
          Affymetrix HT Mouse Genome 430A Array 1432863_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 529369 - -
          ABI Mouse Genome Survey Microarray 662508 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102680088 - -
          MouseWG-6 v2 Expression BeadChip 6940450 - -
          Illumina mouse-6 v1.0 expression beadchip 6940450 - -
          Illumina mouse-6 v1.0 expression beadchip 102680088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2719346 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2719346 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258441 - -
          mouse-6 v1.1 (Illumina) scl00228731.2_330-S - -
          mouse-6 v1.1 (Illumina) scl18554.3.1_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940450 - -
          Illumina Sentrix mouse-6.v1 bead array 102680088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00228731.2_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102680088 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2719346 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2719346 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline