Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840285_m1
          See other LGALS2 GE Assays ›
          Gene Symbol:
          Lgals2
          Gene Name:
          lectin, galactose-binding, soluble 2
          Gene Aliases:
          2200008F12Rik, AI324147
          Chromosome Location:
          Chr.15: 78850860 - 78855529 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840285_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lgals2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025622.3 NP_079898.2 2 - 3 141 85
          GenBank mRNA AK007364.1 - 2 - 3 138 85
          AK008423.1 - 2 - 3 142 85
          AK008607.1 - 2 - 3 143 85
          AK008636.1 - 2 - 3 142 85
          AK008648.1 - 2 - 3 142 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001849.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lgals2

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Process(es)

          galactoside binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AK008607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AK008636.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AK008648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AW456488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 NM_025622.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AK008607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AK008636.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AK008648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 AW456488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563375 NM_025622.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768633 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135791 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AK008607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AK008636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AK008648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AW456488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 NM_025622.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AK008607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AK008636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AK008648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 AW456488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P563375 NM_025622.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30362763 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135791 - -
          Whole Mouse Genome G4122A A_52_P563375 AK008607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AK008636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AK008648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AW456488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 NM_025622.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AK008607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AK008636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AK008648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 AW456488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563375 NM_025622.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110065 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12038520 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12038519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12038516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11907177 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12038517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30362815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11896133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12216551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12138570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12267773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11934089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12122549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12434385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11945108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12084056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12084027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12284748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12247936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11865033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12038517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12070239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11907178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12141743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11896132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12084065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11960075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30362749 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430428 - -
          Affymetrix HT MG-430 PM Array Plate 1417079_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1417078_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430425 - -
          Mouse Genome 430 2.0 Array 1417079_s_at - -
          Mouse Genome 430 2.0 Array 1417078_PM_at - -
          Mouse Genome 430 2.0 Array 1417079_PM_s_at - -
          Mouse Genome 430 2.0 Array 1417078_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107753_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107753_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500014449.mm.1 NM_025622.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014449.mm.1 NM_025622.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014449.mm.1 TC1500001849.mm.1 MTA TC
          Mouse Expression Set 430 1417079_PM_s_at - -
          Mouse Expression Set 430 1417078_at - -
          Mouse Expression Set 430 1417078_PM_at - -
          Mouse Expression Set 430 1417079_s_at - -
          Murine Genome U74v2 Set 160221_PM_at - -
          Murine Genome U74v2 Set 160221_at - -
          Mouse Genome 430A 2.0 Array 1417078_PM_at - -
          Mouse Genome 430A 2.0 Array 1417079_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1417079_s_at - -
          Mouse Genome 430A 2.0 Array 1417078_at - -
          Affymetrix HT Mouse Genome 430A Array 1417078_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417079_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417078_at - -
          Affymetrix HT Mouse Genome 430A Array 1417079_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 604245 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3610487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2641371 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2641371 - -
          mouse-6 v1.1 (Illumina) scl46981.3.21_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610487 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46981.3.21_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2641371 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2641371 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline