Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840775_g1
          See other GHDC GE Assays ›
          Gene Symbol:
          Ghdc
          Gene Name:
          GH3 domain containing
          Gene Aliases:
          D11Lgp1e
          Chromosome Location:
          Chr.11: 100766026 - 100770982 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840775_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ghdc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031871.1 NP_114077.1 8 - 9 1470 81
          GenBank mRNA AF316996.1 - 8 - 9 1470 81
          AK143922.1 - 8 - 9 1439 81
          AK161571.1 - 5 - 6 618 81
          BC005639.1 - 8 - 9 1451 3256858 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003785.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ghdc

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AK161571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 BC005639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AF316996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AK135322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AK143922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AK161571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 BC005639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AF316996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AK135322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P192847 AK143922.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322870 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169100 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AK161571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 BC005639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AF316996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AK135322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AK143922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AK161571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 BC005639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AF316996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AK135322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P192847 AK143922.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28070073 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169100 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07464725 - -
          Whole Mouse Genome G4122A A_52_P629151 BC005639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629151 AF316996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629151 AK135322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629151 AK143922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629151 NM_031871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AK161571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 BC005639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AF316996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AK135322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AK143922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AK161571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 BC005639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AF316996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AK135322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192847 AK143922.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122600 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09625158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09625156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28070019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09609516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09139984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09179778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08990699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09656420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09625160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09446503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09438828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09446500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28070042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391278 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391280 - -
          Affymetrix HT MG-430 PM Array Plate 1419502_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391277 - -
          Mouse Genome 430 2.0 Array 1419502_at - -
          Mouse Genome 430 2.0 Array 1419502_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80860_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80860_at - -
          Affymetrix Mouse Clariom S Array PSR1100034895.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034895.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034895.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034902.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034902.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034902.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034903.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034903.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034903.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034894.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034894.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034894.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034904.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034904.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034904.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034896.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034896.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034896.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034899.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034899.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034899.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034897.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034897.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034897.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034900.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034900.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034900.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034907.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034907.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034907.mm.1 TC1100003785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034905.mm.1 NM_031871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034905.mm.1 NM_031871.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034905.mm.1 TC1100003785.mm.1 MTA TC
          Mouse Expression Set 430 1419502_PM_at - -
          Mouse Expression Set 430 1419502_at - -
          Mouse Genome 430A 2.0 Array 1419502_at - -
          Mouse Genome 430A 2.0 Array 1419502_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419502_at - -
          Affymetrix HT Mouse Genome 430A Array 1419502_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 585623 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 610093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610763 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610763 - -
          mouse-6 v1.1 (Illumina) scl39549.9.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 610093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39549.9.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862465 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862465 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline