Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00841037_m1
          See other CRYGN GE Assays ›
          Gene Symbol:
          Crygn
          Gene Name:
          crystallin, gamma N
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 24751002 - 24757843 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00841037_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crygn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153076.2 NP_694716.2 3 - 4 416 62
          GenBank mRNA AF445456.1 - 3 - 4 416 62
          BC120913.1 - 3 - 4 416 40091405 62
          BC120914.1 - 2 - 3 405 62
          CF739670.1 - 2 - 3 400 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002092.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crygn

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          structural constituent of eye lens

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23543724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23543723 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23543722 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975295 NM_153076.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975295 AF445456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975295 BC120913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975295 BC120914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975295 CF739670.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6946 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160743 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975295 NM_153076.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975295 AF445456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975295 BC120913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975295 BC120914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975295 CF739670.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353814 NM_153076.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353814 AF445456.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353814 BC120913.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353814 BC120914.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353814 CF739670.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23543763 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05507873 - -
          Whole Mouse Genome G4122A A_51_P353814 NM_153076.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353814 AF445456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353814 BC120913.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353814 BC120914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353814 CF739670.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109403 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160743 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971157 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971156 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04971162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23543724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23543722 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23543723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23543775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04124763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04706259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04245142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04706252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04706247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04942340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04971156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04123865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04706255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04635847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04677660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04635842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04176085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04971151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23543706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10528687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528690 - -
          Affymetrix HT MG-430 PM Array Plate 1425286_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10528686 - -
          Mouse Genome 430 2.0 Array 1425286_PM_at - -
          Mouse Genome 430 2.0 Array 1425286_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214301_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214301_at - -
          Affymetrix Mouse Clariom S Array PSR0500019074.mm.1 TC0500002092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019074.mm.1 NM_153076.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019074.mm.1 NM_153076.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019076.mm.1 TC0500002092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019076.mm.1 NM_153076.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019076.mm.1 NM_153076.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019077.mm.1 TC0500002092.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019077.mm.1 NM_153076.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019077.mm.1 NM_153076.2 Refseq
          Mouse Expression Set 430 1425286_PM_at - -
          Mouse Expression Set 430 1425286_at - -
          Mouse Genome 430A 2.0 Array 1425286_PM_at - -
          Mouse Genome 430A 2.0 Array 1425286_at - -
          Affymetrix HT Mouse Genome 430A Array 1425286_at - -
          Affymetrix HT Mouse Genome 430A Array 1425286_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 477259 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2350403 - -
          Illumina mouse-6 v1.0 expression beadchip 2350403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774572 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774572 - -
          mouse-6 v1.1 (Illumina) scl26794.4.1_114-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26794.4.1_114-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3102770 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3029509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3029509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3102770 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774572 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0