| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10417502 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417380 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417334 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417378 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417447 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417448 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417463 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417489 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417372 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417456 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417352 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417389 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417454 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417457 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417451 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417450 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417428 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417466 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417453 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417449 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417455 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417464 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10417419 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417286 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417485 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417326 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417452 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417446 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417421 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417461 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417415 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417373 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10417501 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1400000493.mm.1 |
NM_175393.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000493.mm.1 |
NM_175393.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000493.mm.1 |
TC1400000038.mm.1 |
MTA TC |