| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10356252 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10347930 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582872 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356250 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356290 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356287 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582876 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10347929 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582870 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10534910 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582875 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356282 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10534911 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356283 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356279 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582869 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356253 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356284 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582871 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356249 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356289 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356281 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356286 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356285 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356280 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10356251 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1456493_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1429562_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1429563_PM_x_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10582868 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10347928 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10356278 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10534909 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10582874 |
- |
- |
| Mouse Genome 430 2.0 Array |
1429563_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429563_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1425078_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447388_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429562_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1425078_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447388_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429562_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0100022049.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022049.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022049.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022049.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022049.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022052.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022052.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022052.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022052.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022052.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022054.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022054.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022054.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022054.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022054.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022057.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022057.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022057.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022057.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022057.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022046.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022046.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022046.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022046.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022046.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022047.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022047.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022047.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022047.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022047.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022050.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022050.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022050.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022050.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022050.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022053.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022053.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022053.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022053.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022053.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022048.mm.1 |
NM_030194.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022048.mm.1 |
NM_030194.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022048.mm.1 |
NM_175397.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022048.mm.1 |
NM_175397.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022048.mm.1 |
TC0100002725.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1429562_PM_at |
- |
- |
| Mouse Expression Set 430 |
1429562_at |
- |
- |
| Mouse Expression Set 430 |
1429563_x_at |
- |
- |
| Mouse Expression Set 430 |
1429563_PM_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1429563_PM_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1429563_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1429562_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1429562_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1429562_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1429563_PM_x_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1429563_x_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1429562_PM_at |
- |
- |