Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00841632_m1
          See other SKINT9 GE Assays ›
          Gene Symbol:
          Skint9
          Gene Name:
          selection and upkeep of intraepithelial T cells 9
          Gene Aliases:
          A030013N09Rik, Skint-9
          Chromosome Location:
          Chr.4: 112385969 - 112433985 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00841632_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Skint9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177864.2 NP_808532.1 5 - 6 707 94
          GenBank mRNA AK037244.1 - 5 - 6 707 94
          EF494907.1 - 5 - 6 699 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003290.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Skint9

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157388 NM_177864.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157388 AK037244.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839958 BC139813.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839958 NM_146922.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839958 NM_147024.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33330 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173459 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118751 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157388 NM_177864.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157388 AK037244.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23214380 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00962261 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173459 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05361995 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00962255 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118751 - -
          Whole Mouse Genome G4122A A_51_P495747 EF494907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495747 NM_177864.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495747 AK037244.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119740 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116514 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118751 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05361995 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173459 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03725631 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03725642 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03878414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03725639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23214287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03475231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03878414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23214372 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515137 - -
          Affymetrix Mouse Gene 1.1 ST Array 10515134 - -
          Affymetrix Mouse Clariom S Array PSR0400026192.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026192.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026192.mm.1 NM_177864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026189.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026189.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026189.mm.1 NM_177864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026186.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026186.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026186.mm.1 NM_177864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026187.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026187.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026187.mm.1 NM_177864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026188.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026188.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026188.mm.1 NM_177864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026190.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026190.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026190.mm.1 NM_177864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026191.mm.1 TC0400003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026191.mm.1 NM_177864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026191.mm.1 NM_177864.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704450 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 450184 - -
          Illumina mouse-6 v1.0 expression beadchip 450184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000450184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710079 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710079 - -
          mouse-6 v1.1 (Illumina) scl23983.7.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 450184 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23983.7.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 450184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2979289 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2979289 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0