Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00842045_g1
          See other DEFA20 GE Assays ›
          Gene Symbol:
          Defa20 Defa2 Gm15308 Gm21002
          Gene Name:
          defensin, alpha, 20
          defensin, alpha, 2
          predicted gene 15308
          predicted gene, 21002
          Gene Aliases:
          2010016F14Rik, Defcr20
          CR2, Defcr-2, Defcr2
          ENSMUSG00000065957, OTTMUSG00000019860
          -
          Chromosome Location:
          Chr.8: 21509258 - 21510237 on Build GRCm38
          Chromosome Location:
          Chr.8: 21378517 - 21379495 on Build GRCm38
          Chromosome Location:
          Chr.8: 21529030 - 21530012 on Build GRCm38
          Chromosome Location:
          Chr.8: 21391790 - 21418342 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          Assay ID Mm00842045_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defa20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183268.4 NP_899091.2 1 - 2 220 146
          GenBank mRNA AK008107.1 - 1 - 2 221 146
          AK008275.1 - 1 - 2 221 146
          Gene Symbol: Defa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195634.2 NP_001182563.2 1 - 2 220 146
          GenBank mRNA AK008459.1 - 1 - 2 223 146
          Gene Symbol: Gm15308
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177521.1 NP_001170992.1 222 146
          Gene Symbol: Gm21002
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270555.1 NP_001257484.1 222 146
          GenBank mRNA AY746425.1 - 177 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defa20
          Click here to find siRNAs for Gene Defa2
          Click here to find siRNAs for Gene Gm15308
          Click here to find siRNAs for Gene Gm21002

          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antibacterial humoral response
          response to drug
          defense response to bacterium
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          cellular response to lipopolysaccharide
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961408 NM_001177521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961408 AK008377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961408 AK008459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961408 AY746425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961408 DQ103703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961407 NM_001177521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961407 AK008377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961407 AK008459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961407 AY746425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961407 DQ103703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961407 NM_001177521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961407 AK008377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961407 AK008459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961407 AY746425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961407 DQ103703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961408 NM_001177521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961408 AK008377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961408 AK008459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961408 AY746425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961408 DQ103703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249336 NM_001177521.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249336 DQ103703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249336 NM_183268.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249336 AK008107.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249336 AK008275.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25706592 - -
          Whole Mouse Genome G4122A A_52_P249336 NM_001177521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249336 DQ103703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249336 NM_183268.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249336 AK008107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249336 AK008275.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06807227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07032216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07006212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07405865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25706535 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10570709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570710 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570706 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570711 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570711 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570693 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570706 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570693 - -
          Affymetrix Mouse Clariom S Array PSR0800001860.mm.1 NM_001195634.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001860.mm.1 TC0800000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001861.mm.1 NM_001195634.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001861.mm.1 TC0800000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001866.mm.1 NM_001270555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001866.mm.1 TC0800000235.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001890.mm.1 NM_001177521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001890.mm.1 NM_001177521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001890.mm.1 TC0800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001892.mm.1 NM_001177521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001892.mm.1 NM_001177521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001892.mm.1 TC0800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001863.mm.1 NM_001270555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001863.mm.1 TC0800000235.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001864.mm.1 NM_001270555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001864.mm.1 TC0800000235.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001893.mm.1 NM_001177521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001893.mm.1 NM_001177521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001893.mm.1 TC0800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001885.mm.1 NM_183268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001885.mm.1 TC0800000242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001859.mm.1 NM_001195634.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001859.mm.1 TC0800000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001887.mm.1 NM_183268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001887.mm.1 TC0800000242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001891.mm.1 NM_001177521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001891.mm.1 NM_001177521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001891.mm.1 TC0800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001862.mm.1 NM_001195634.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001862.mm.1 TC0800000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001867.mm.1 NM_001270555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001867.mm.1 TC0800000235.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001886.mm.1 NM_183268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001886.mm.1 TC0800000242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001888.mm.1 NM_183268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001888.mm.1 TC0800000242.mm.1 MTA TC
          Murine Genome U74v2 Set 92812_f_at - -
          Murine Genome U74v2 Set 92812_f_at - -
          Murine Genome U74v2 Set 92812_PM_f_at - -
          Murine Genome U74v2 Set 92812_PM_f_at - -
          Murine Genome U74v2 Set 165407_f_at - -
          Murine Genome U74v2 Set 165407_PM_f_at - -
          Murine Genome U74v2 Set 165492_PM_at - -
          Murine Genome U74v2 Set 165492_at - -
          Murine Genome U74v2 Set 165407_PM_f_at - -
          Murine Genome U74v2 Set 165407_f_at - -
          Murine Genome U74v2 Set 165492_at - -
          Murine Genome U74v2 Set 165492_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5080575 - -
          Illumina mouse-6 v1.0 expression beadchip 102900750 - -
          Illumina mouse-6 v1.0 expression beadchip 4670022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230099 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2675560 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2675560 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234291 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230099 - -
          mouse-6 v1.1 (Illumina) scl013238.1_31-S - -
          mouse-6 v1.1 (Illumina) scl0068009.1_159-S - -
          mouse-6 v1.1 (Illumina) GI_34304055-S - -
          Illumina Sentrix mouse-6.v1 bead array 102900750 - -
          Illumina Sentrix mouse-6.v1 bead array 4670022 - -
          Illumina Sentrix mouse-6.v1 bead array 5080575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013238.1_31-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0068009.1_159-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670022 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102900750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2675560 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2739019 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230099 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2675560 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2739019 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0