Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00842897_m1
          See other AA792892 GE Assays ›
          Gene Symbol:
          AA792892
          Gene Name:
          expressed sequence AA792892
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 94339763 - 94384334 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00842897_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AA792892
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178894.4 NP_849225.2 2 - 3 406 158
          GenBank mRNA AK053948.1 - 361 158
          AK087752.1 - 430 158
          BC120516.1 - 2 - 3 406 40129439 158
          BC125434.1 - 2 - 3 406 40129949 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AA792892

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965649 AK087752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965649 BC120516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965649 BC125434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965649 NM_178894.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965649 AK053948.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965649 AK087752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965649 BC120516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965649 BC125434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965649 NM_178894.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965649 AK053948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P398730 BC083101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P398730 AK087752.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106849 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536043 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531505 - -
          Affymetrix Mouse Gene 1.1 ST Array 10536029 - -
          Affymetrix Mouse Gene 1.1 ST Array 10536041 - -
          Affymetrix Mouse Clariom S Array PSR0500007799.mm.1 TC0500000903.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007799.mm.1 NM_178894.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007799.mm.1 NM_178894.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007800.mm.1 TC0500000903.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007800.mm.1 NM_178894.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007800.mm.1 NM_178894.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007798.mm.1 TC0500000903.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007798.mm.1 NM_178894.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007798.mm.1 NM_178894.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 755678 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103170465 - -
          Illumina mouse-6 v1.0 expression beadchip 103170465 - -
          Illumina mouse-6 v1.0 expression beadchip 4010402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010402 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235804 - -
          mouse-6 v1.1 (Illumina) scl0100554.1_330-S - -
          mouse-6 v1.1 (Illumina) GI_31342261-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010402 - -
          Illumina Sentrix mouse-6.v1 bead array 103170465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0100554.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103170465 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2984591 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2984591 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline