| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10533004 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10533006 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10533005 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1420750_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10533003 |
- |
- |
| Mouse Genome 430 2.0 Array |
1420750_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420750_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500027621.mm.1 |
TC0500003103.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027621.mm.1 |
NR_027810.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027621.mm.1 |
NR_027810.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027623.mm.1 |
TC0500003103.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027623.mm.1 |
NR_027810.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027623.mm.1 |
NR_027810.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027622.mm.1 |
TC0500003103.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027622.mm.1 |
NR_027810.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027622.mm.1 |
NR_027810.1 |
Refseq |
| Mouse Expression Set 430 |
1420750_at |
- |
- |
| Mouse Expression Set 430 |
1420750_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420750_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420750_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1420750_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1420750_at |
- |
- |