Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00843581_s1
          See other CXX1A GE Assays ›
          Gene Symbol:
          Cxx1a
          Gene Name:
          CAAX box 1A
          Gene Aliases:
          1110012O05Rik, Mar8.2A/B, Mart8b
          Chromosome Location:
          Chr.X: 53642488 - 53643763 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00843581_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxx1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024170.2 NP_077132.1 1 825 179
          GenBank mRNA AK002323.1 - 1 761 179
          BC003436.1 - 1 744 2647372 179

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985591 NM_001018063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985591 AK003646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985591 BC029838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985591 NM_024170.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985591 AK002323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985591 NM_001018063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985591 AK003646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985591 BC029838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985591 NM_024170.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985591 AK002323.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14660043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14660046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14660044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14877501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14902392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14596268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14972297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14812467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14877496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14877501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14902386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14852040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14892712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14972298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14947959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10604638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604635 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604637 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604633 - -
          Mouse Genome 430 2.0 Array 1423816_PM_at - -
          Mouse Genome 430 2.0 Array 1423816_at - -
          Mouse Expression Set 430 1423816_at - -
          Mouse Expression Set 430 1423816_PM_at - -
          Murine Genome U74v2 Set 99609_PM_at - -
          Murine Genome U74v2 Set 99609_at - -
          Murine Genome U74v2 Set 166567_f_at - -
          Murine Genome U74v2 Set 166567_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1423816_PM_at - -
          Mouse Genome 430A 2.0 Array 1423816_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 921644 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1050685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050685 - -
          mouse-6 v1.1 (Illumina) scl066158.1_320-S - -
          Illumina Sentrix mouse-6.v1 bead array 1050685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066158.1_320-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2952779 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2952779 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2595200 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066158.1_320-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline