Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844026_s1
          See other MAGEL2 GE Assays ›
          Gene Symbol:
          Magel2
          Gene Name:
          melanoma antigen, family L, 2
          Gene Aliases:
          Mage-l2, NDNL1, nM15, ns7
          Chromosome Location:
          Chr.7: 62376979 - 62381640 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844026_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Magel2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013779.2 NP_038807.4 1 4435 93
          GenBank mRNA AJ243608.1 - 1 1950 93
          AK086725.1 - 1 4431 93
          BC054763.1 - 1 1727 93
          BC062913.1 - 3 3940 93
          BC071222.1 - 1 2213 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Magel2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          Arp2/3 complex-mediated actin nucleation
          retrograde transport, endosome to Golgi
          regulation of circadian rhythm
          negative regulation of transcription, DNA-templated
          rhythmic process
          positive regulation of actin nucleation
          protein K63-linked ubiquitination
          ubiquitin-protein transferase activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003251 AJ243608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003251 AK086725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003251 BC054763.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003251 BC062913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003251 BC071222.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165384 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149329 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003251 AJ243608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003251 AK086725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003251 BC054763.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003251 BC062913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003251 BC071222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286798 AJ243608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286798 AK082944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286798 AK086725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286798 BC054763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286798 BC062913.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06007341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06007360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06018435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06269416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06269427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06477261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06154317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06154315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05931972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06269391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25168819 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10553837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553836 - -
          Affymetrix HT MG-430 PM Array Plate 1417217_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10553835 - -
          Mouse Genome 430 2.0 Array 1417217_PM_at - -
          Mouse Genome 430 2.0 Array 1417217_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27385_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27385_at - -
          Affymetrix Mouse Clariom S Array PSR0700008763.mm.1 TC0700000962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008763.mm.1 NM_013779.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008763.mm.1 NM_013779.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008761.mm.1 TC0700000962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008761.mm.1 NM_013779.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008761.mm.1 NM_013779.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008762.mm.1 TC0700000962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008762.mm.1 NM_013779.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008762.mm.1 NM_013779.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008760.mm.1 TC0700000962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008760.mm.1 NM_013779.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008760.mm.1 NM_013779.2 Refseq
          Mouse Expression Set 430 1417217_PM_at - -
          Mouse Expression Set 430 1417217_at - -
          Murine Genome U74v2 Set 92681_at - -
          Murine Genome U74v2 Set 92681_PM_at - -
          Mouse Genome 430A 2.0 Array 1417217_PM_at - -
          Mouse Genome 430A 2.0 Array 1417217_at - -
          Affymetrix HT Mouse Genome 430A Array 1417217_at - -
          Affymetrix HT Mouse Genome 430A Array 1417217_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 562819 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7050619 - -
          Illumina mouse-6 v1.0 expression beadchip 7050619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694539 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694539 - -
          mouse-6 v1.1 (Illumina) scl32591.1_99-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32591.1_99-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2694539 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694539 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline