Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844196_s1
          See other 4933414I15RIK GE Assays ›
          Gene Symbol:
          4933414I15Rik
          Gene Name:
          RIKEN cDNA 4933414I15 gene
          Gene Aliases:
          4932442I06Rik, 4933401E19Rik, ENSMUSG00000051192
          Chromosome Location:
          Chr.11: 50940819 - 50943833 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844196_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933414I15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016566.1 - 1 2636 129
          AK016600.1 - 2 1797 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382139 AK016600.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382139 AK016813.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382139 AK016566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382139 AK016600.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382139 AK016813.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382139 AK016566.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791414 - -
          Whole Mouse Genome G4122A A_52_P382139 AK016600.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382139 AK016813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382139 AK016566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382139 AK016600.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382139 AK016813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382139 AK016566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550062 AK016600.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550062 AK016566.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132702 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431470_PM_a_at - -
          Mouse Genome 430 2.0 Array 1431470_a_at - -
          Mouse Genome 430 2.0 Array 1431470_PM_a_at - -
          Mouse Expression Set 430 1431470_a_at - -
          Mouse Expression Set 430 1431470_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1431470_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1431470_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 795944 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6510309 - -
          Illumina mouse-6 v1.0 expression beadchip 6510309 - -
          Illumina mouse-6 v1.0 expression beadchip 100380035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258651 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258651 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2545478 - -
          mouse-6 v1.1 (Illumina) scl074408.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 100380035 - -
          Illumina Sentrix mouse-6.v1 bead array 6510309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl074408.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100380035 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510309 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2545478 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766213 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline