Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844278_s1
          See other VMN1R148 GE Assays ›
          Gene Symbol:
          Vmn1r148
          Gene Name:
          vomeronasal 1 receptor 148
          Gene Aliases:
          V1rd14, V3R8
          Chromosome Location:
          Chr.7: 22412231 - 22414669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844278_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r148
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030736.2 NP_109661.1 1 2250 113
          GenBank mRNA AF324872.1 - 1 2252 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r148

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056450 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163904 NM_001166847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163904 NM_001166845.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163904 NM_001166844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163904 NM_001166757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163904 NM_001166754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056450 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P170316 NM_030736.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P170316 AF324872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170316 NM_030736.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170316 AF324872.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119393 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110443 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115610 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560743 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550783 - -
          Affymetrix HT MG-430 PM Array Plate 1450315_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560795 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550765 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560752 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550778 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560754 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550782 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550786 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560767 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550760 - -
          Mouse Genome 430 2.0 Array 1450315_at - -
          Mouse Genome 430 2.0 Array 1450315_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002555.mm.1 TC0700000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002555.mm.1 NM_030736.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002555.mm.1 NM_030736.2 Refseq
          Mouse Expression Set 430 1450315_PM_at - -
          Mouse Expression Set 430 1450315_at - -
          Mouse Genome 430A 2.0 Array 1450315_PM_at - -
          Mouse Genome 430A 2.0 Array 1450315_at - -
          Affymetrix HT Mouse Genome 430A Array 1450315_at - -
          Affymetrix HT Mouse Genome 430A Array 1450315_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 403400 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4230717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2473597 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2473597 - -
          mouse-6 v1.1 (Illumina) scl081011.3_270-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl081011.3_270-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2473597 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2473597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline