Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844303_s1
          See other PCDHB1 GE Assays ›
          Gene Symbol:
          Pcdhb1
          Gene Name:
          protocadherin beta 1
          Gene Aliases:
          PcdhbA
          Chromosome Location:
          Chr.18: 37264998 - 37267454 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844303_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053126.1 NP_444356.1 1 2408 140
          GenBank mRNA AY013770.1 - 1 2408 140
          BC132112.1 - 1 2468 40130389 140
          BC132296.1 - 1 2468 40130573 140

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb1

          Panther Classification:

          Molecular Function -

          cadherin

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173288 BC132296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173288 AY013770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173288 BC132112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173288 NM_053126.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56706 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127639 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173288 BC132296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173288 AY013770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173288 BC132112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173288 NM_053126.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468912 BC132296.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468912 AY013770.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468912 BC132112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468912 NM_053126.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627537 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127639 - -
          Whole Mouse Genome G4122A A_51_P468912 BC132296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468912 AY013770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468912 BC132112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468912 NM_053126.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104120 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13888933 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13948192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13888950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13888940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13888955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13680211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13888962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13905041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13905043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13945123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13680213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14103707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13609791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455049 - -
          Affymetrix HT MG-430 PM Array Plate 1421631_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455048 - -
          Mouse Genome 430 2.0 Array 1421631_at - -
          Mouse Genome 430 2.0 Array 1421631_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93872_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93872_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800002809.mm.1 TC1800000355.mm.1 MTA TC
          Mouse Expression Set 430 1421631_PM_at - -
          Mouse Expression Set 430 1421631_at - -
          Mouse Genome 430A 2.0 Array 1421631_at - -
          Mouse Genome 430A 2.0 Array 1421631_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421631_at - -
          Affymetrix HT Mouse Genome 430A Array 1421631_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 430063 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2260706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724088 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724088 - -
          mouse-6 v1.1 (Illumina) scl52074.1.1_285-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52074.1.1_285-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2724088 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2724088 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline