Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844610_s1
          See other KIR3DL2 GE Assays ›
          Gene Symbol:
          Kir3dl2
          Gene Name:
          killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2
          Gene Aliases:
          Kirl2
          Chromosome Location:
          Chr.X: 136447841 - 136544052 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844610_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kir3dl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177748.2 NP_808416.1 9 1650 117
          GenBank mRNA AF548540.1 - 9 1650 117

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kir3dl2

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027920 BC132015.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027920 AF548541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027920 AK046171.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027920 AY530784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027920 BC132013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2089313 NM_177748.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2089313 AF548540.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138789 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027920 BC132015.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027920 AF548541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027920 AK046171.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027920 AY530784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027920 BC132013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2089313 NM_177748.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2089313 AF548540.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430097 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03759863 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430138 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430156 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430085 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430126 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430113 - -
          Whole Mouse Genome G4122A A_51_P250860 BC132015.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250860 AF548541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250860 AK046171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250860 AY530784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250860 BC132013.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136942 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103648 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116177 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09430113 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03759860 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09430085 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09430146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14674262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15174430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681163 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606875 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606870 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3830093 - -
          Illumina mouse-6 v1.0 expression beadchip 1770593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2761690 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2761690 - -
          mouse-6 v1.1 (Illumina) scl0245616.3_12-S - -
          mouse-6 v1.1 (Illumina) scl0245615.3_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770593 - -
          Illumina Sentrix mouse-6.v1 bead array 3830093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0245616.3_12-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0245615.3_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830093 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2761691 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223916 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2761691 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223916 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0