Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844994_s1
          See other OBOX3 GE Assays ›
          Gene Symbol:
          Obox3
          Gene Name:
          oocyte specific homeobox 3
          Gene Aliases:
          Ohx
          Chromosome Location:
          Chr.7: 15625306 - 15639777 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844994_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Obox3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145707.3 NP_663753.3 5 965 148
          GenBank mRNA AK139750.1 - 5 966 148
          AY094981.1 - 1355 148
          BC119564.1 - 1 1818 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Obox3

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131659 BC119564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131659 NM_145707.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131659 AK139750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052076 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015012 AY094981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015012 BC119564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015012 NM_145707.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015012 AK139750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052076 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015012 AY094981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015012 BC119564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015012 NM_145707.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015012 AK139750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131659 BC119564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131659 NM_145707.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131659 AK139750.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481448 AY094981.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128215 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128184 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128230 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128201 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01480913 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06128168 - -
          Whole Mouse Genome G4122A A_51_P481448 AY094981.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06128168 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06128184 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06128231 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06128217 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01480913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27838580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27838146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27838549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27838146 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550198 - -
          Affymetrix HT MG-430 PM Array Plate 1452634_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560160 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560153 - -
          Mouse Genome 430 2.0 Array 1452634_PM_at - -
          Mouse Genome 430 2.0 Array 1452634_at - -
          Affymetrix Mouse Clariom S Array PSR0700021173.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021173.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021173.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021174.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021174.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021174.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021160.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021160.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021160.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021165.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021165.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021165.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021170.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021170.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021170.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021176.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021176.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021176.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021164.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021164.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021164.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021163.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021163.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021163.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021168.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021168.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021168.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021166.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021166.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021166.mm.1 NM_145707.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021161.mm.1 TC0700002407.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021161.mm.1 NM_145707.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021161.mm.1 NM_145707.3 Refseq
          Mouse Expression Set 430 1452634_at - -
          Mouse Expression Set 430 1452634_PM_at - -
          Mouse Genome 430A 2.0 Array 1452634_at - -
          Mouse Genome 430A 2.0 Array 1452634_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452634_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452634_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 801090 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 540673 - -
          Illumina mouse-6 v1.0 expression beadchip 540673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540673 - -
          mouse-6 v1.1 (Illumina) scl0246791.1_221-S - -
          Illumina Sentrix mouse-6.v1 bead array 540673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0246791.1_221-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3147577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3069117 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3147577 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2751259 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3069117 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline