| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10520272 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1453655_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10520271 |
- |
- |
| Mouse Genome 430 2.0 Array |
1453655_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453655_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500001755.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001755.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001755.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001751.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001751.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001751.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001750.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001750.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001750.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001754.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001754.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001754.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001753.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001753.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001753.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001757.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001757.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001757.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001749.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001749.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001749.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001756.mm.1 |
NR_027851.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001756.mm.1 |
NR_027851.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001756.mm.1 |
TC0500000226.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1453655_PM_at |
- |
- |
| Mouse Expression Set 430 |
1453655_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453655_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453655_PM_at |
- |
- |