Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00847785_g1
          See other OLFR900 GE Assays ›
          Gene Symbol:
          Olfr900
          Gene Name:
          olfactory receptor 900
          Gene Aliases:
          GA_x6K02T2PVTD-32071567-32072508, MOR170-13, MOR170-2
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00847785_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr900
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146874.1 NP_667085.1 1 357 156
          GenBank mRNA BC106986.2 - 1 357 40043276 156

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr900

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P423104 NM_146874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P423104 BC106986.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P423104 NM_146874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P423104 BC106986.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P423104 NM_146874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P423104 BC106986.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P423104 NM_146874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P423104 BC106986.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P423104 NM_146874.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P423104 BC106986.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P423104 NM_146874.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P423104 BC106986.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434527 - -
          Whole Mouse Genome G4122A A_52_P423104 NM_146874.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423104 BC106986.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423104 NM_146874.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423104 BC106986.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730032 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08045514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584400 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584403 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584399 - -
          Affymetrix Mouse Clariom S Array PSR0900002793.mm.1 NM_207553.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002793.mm.1 NM_207553.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002793.mm.1 TC0900000397.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 885664 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 580739 - -
          Illumina mouse-6 v1.0 expression beadchip 580739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2758599 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2758599 - -
          mouse-6 v1.1 (Illumina) scl00258874.1_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 580739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258874.1_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2913877 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2758599 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2758599 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913877 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline