Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00848629_sH
          See other OLFR826 GE Assays ›
          Gene Symbol:
          Olfr826
          Gene Name:
          olfactory receptor 826
          Gene Aliases:
          MOR210-2
          Chromosome Location:
          Chr.10: 130179937 - 130184162 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00848629_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr826
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146676.1 NP_666887.1 1 674 164

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003205.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr826

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P385526 NM_146676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P385526 NM_146676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098895 NM_146676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098895 NM_146677.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098895 BC119054.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098895 BC119056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P385526 NM_146676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P385526 NM_146676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098895 NM_146676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098895 NM_146677.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098895 BC119054.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098895 BC119056.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P385526 NM_146676.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P385526 NM_146676.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548637 - -
          Whole Mouse Genome G4122A A_52_P385526 NM_146676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P385526 NM_146676.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103014 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08757054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08757063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08468873 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08468875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08468872 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08757067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548609 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373677 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373676 - -
          Affymetrix Mouse Clariom S Array PSR1000023989.mm.1 NM_146676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023989.mm.1 NM_146676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023989.mm.1 TC1000003205.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5080142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2588637 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588637 - -
          mouse-6 v1.1 (Illumina) scl5152.1.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5152.1.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2588637 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2588637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline