Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00849899_g1
          See other RPL36A GE Assays ›
          Gene Symbol:
          Rpl36a
          Gene Name:
          ribosomal protein L36A
          Gene Aliases:
          L44L, Rpl44
          Chromosome Location:
          Chr.X: 134585654 - 134588062 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00849899_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl36a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019865.5 NP_063918.1 4 - 5 363 158
          GenBank mRNA AK012502.1 - 4 - 5 333 158
          AK170127.1 - 4 - 5 332 158
          AV453514.1 - 4 - 5 318 158
          BC027515.1 - 4 - 5 318 1383212 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl36a

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672686 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672689 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672685 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672687 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019019 AV453514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019019 NM_019865.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019019 AK012502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102816 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881850 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120282 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102816 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019019 AV453514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019019 NM_019865.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019019 AK012502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956707 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283920 AK170127.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283920 AV453514.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283920 BC027515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283920 NM_019865.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283920 AK012502.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P481880 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P197926 AK170127.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P197926 AV453514.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P197926 BC027515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P197926 NM_019865.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P197926 AK012502.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672698 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120282 - -
          Whole Mouse Genome G4122A A_51_P283920 AK170127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283920 AV453514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283920 BC027515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283920 NM_019865.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283920 AK012502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P197926 AK170127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P197926 AV453514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P197926 BC027515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P197926 NM_019865.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P197926 AK012502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P481880 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131024 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118169 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14998770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14998771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672689 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672686 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672687 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14998771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14998776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14998775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14998762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14697087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14998769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601757 - -
          Affymetrix HT MG-430 PM Array Plate 1416807_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10601755 - -
          Affymetrix Mouse Gene 1.1 ST Array 10496735 - -
          Affymetrix Mouse Gene 1.1 ST Array 10421972 - -
          Mouse Genome 430 2.0 Array 1416807_PM_at - -
          Mouse Genome 430 2.0 Array 1438926_at - -
          Mouse Genome 430 2.0 Array 1416807_at - -
          Mouse Genome 430 2.0 Array 1438926_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00007989.mm.1 NM_019865.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007989.mm.1 NM_019865.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007989.mm.1 TC0X00001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008003.mm.1 NM_019865.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008003.mm.1 NM_019865.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008003.mm.1 TC0X00001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008005.mm.1 NM_019865.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008005.mm.1 NM_019865.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008005.mm.1 TC0X00001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007997.mm.1 NM_019865.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007997.mm.1 NM_019865.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007997.mm.1 TC0X00001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007999.mm.1 NM_019865.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007999.mm.1 NM_019865.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007999.mm.1 TC0X00001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008002.mm.1 NM_019865.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008002.mm.1 NM_019865.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008002.mm.1 TC0X00001282.mm.1 MTA TC
          Mouse Expression Set 430 1438926_at - -
          Mouse Expression Set 430 1448697_PM_s_at - -
          Mouse Expression Set 430 1416807_at - -
          Mouse Expression Set 430 1416807_PM_at - -
          Mouse Expression Set 430 1448697_s_at - -
          Mouse Expression Set 430 1438926_PM_at - -
          Murine Genome U74v2 Set 160081_at - -
          Murine Genome U74v2 Set 160081_PM_at - -
          Mouse Genome 430A 2.0 Array 1416807_at - -
          Mouse Genome 430A 2.0 Array 1448697_s_at - -
          Mouse Genome 430A 2.0 Array 1438926_PM_at - -
          Mouse Genome 430A 2.0 Array 1416807_PM_at - -
          Mouse Genome 430A 2.0 Array 1438926_at - -
          Mouse Genome 430A 2.0 Array 1448697_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 891768 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220435 - -
          Illumina mouse-6 v1.0 expression beadchip 5220435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219646 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219646 - -
          mouse-6 v1.1 (Illumina) scl019982.5_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019982.5_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753219 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2753219 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019982.5_48-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline