Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850060_s1
          See other RPS9 GE Assays ›
          Gene Symbol:
          Rps9
          Gene Name:
          ribosomal protein S9
          Gene Aliases:
          3010033P07Rik, AL022771, AL022885
          Chromosome Location:
          Chr.7: 3703953 - 3706899 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00850060_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029767.2 NP_084043.1 3 257 111
          GenBank mRNA AK050591.1 - 3 260 111
          AK081889.1 - 3 261 111
          AK150800.1 - 3 255 111
          AK150988.1 - 3 259 111
          AK151029.1 - 3 255 111
          AK151621.1 - 3 261 111
          BC012491.1 - 2 206 111
          BC031746.1 - 2 247 3494314 111

          Back To Top

          More Information


          Updated Mapping Information

          Based on assay re-evaluation in August 2010 with NCBI transcript and genome annotations (Genome Build NCBIM37.1), this assay is now annotated to be across exon-exon junction.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps9

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          regulation of translation
          positive regulation of cell proliferation
          positive regulation of translational fidelity
          RNA binding
          structural constituent of ribosome
          rRNA binding
          poly(A) RNA binding
          translation regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946918 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946920 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946919 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946921 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946917 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946915 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946916 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047571 BC012491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047571 NM_029767.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047571 AK050591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047571 AK081889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047571 AK150800.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK039648.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK047901.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK140782.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK154588.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK155046.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK155444.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 AK157628.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 BC026483.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729073 NM_146065.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442324 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140535 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047571 BC012491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047571 NM_029767.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047571 AK050591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047571 AK081889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047571 AK150800.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436928 BC012491.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436928 BC031746.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436928 NM_029767.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436928 AK050591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436928 AK081889.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946936 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140535 - -
          Whole Mouse Genome G4122A A_51_P436928 BC012491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436928 BC031746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436928 NM_029767.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436928 AK050591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436928 AK081889.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126490 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140535 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946921 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946916 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946917 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946915 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946920 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946918 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549590 - -
          Affymetrix HT MG-430 PM Array Plate 1426958_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433689_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1434624_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569069 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549588 - -
          Mouse Genome 430 2.0 Array 1434624_PM_x_at - -
          Mouse Genome 430 2.0 Array 1426958_PM_at - -
          Mouse Genome 430 2.0 Array 1433689_s_at - -
          Mouse Genome 430 2.0 Array 1434624_x_at - -
          Mouse Genome 430 2.0 Array 1426958_at - -
          Mouse Genome 430 2.0 Array 1433689_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76846_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76846_at - -
          Affymetrix Mouse Clariom S Array PSR0700000271.mm.1 TC0700000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000271.mm.1 NM_029767.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000271.mm.1 NM_029767.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000265.mm.1 TC0700000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000265.mm.1 NM_029767.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000265.mm.1 NM_029767.2 Refseq
          Mouse Expression Set 430 1433689_s_at - -
          Mouse Expression Set 430 1434624_x_at - -
          Mouse Expression Set 430 1434624_PM_x_at - -
          Mouse Expression Set 430 1426958_at - -
          Mouse Expression Set 430 1433689_PM_s_at - -
          Mouse Expression Set 430 1426958_PM_at - -
          Mouse Genome 430A 2.0 Array 1434624_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1433689_s_at - -
          Mouse Genome 430A 2.0 Array 1426958_PM_at - -
          Mouse Genome 430A 2.0 Array 1433689_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1434624_x_at - -
          Mouse Genome 430A 2.0 Array 1426958_at - -
          Affymetrix HT Mouse Genome 430A Array 1433689_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1433689_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1434624_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426958_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434624_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426958_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 690350 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610021 - -
          Illumina mouse-6 v1.0 expression beadchip 3610021 - -
          Illumina mouse-6 v1.0 expression beadchip 4210397 - -
          Illumina mouse-6 v1.0 expression beadchip 6980131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214918 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1377921 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2588053 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643092 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1377921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214918 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673154 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588053 - -
          mouse-6 v1.1 (Illumina) scl076846.5_166-S - -
          mouse-6 v1.1 (Illumina) scl000275.1_39-S - -
          mouse-6 v1.1 (Illumina) scl0076846.1_142-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610021 - -
          Illumina Sentrix mouse-6.v1 bead array 6980131 - -
          Illumina Sentrix mouse-6.v1 bead array 4210397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl076846.5_166-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0076846.1_142-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000275.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210397 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980131 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214918 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214918 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0076846.1_142-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000275.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl076846.5_166-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline