Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850282_g1
          See other DYNLL1 GE Assays ›
          Gene Symbol:
          Dynll1
          Gene Name:
          dynein light chain LC8-type 1
          Gene Aliases:
          Dlc8, Dnclc1, Pin
          Chromosome Location:
          Chr.5: 115297110 - 115300990 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00850282_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dynll1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019682.4 NP_062656.3 3 575 145
          GenBank mRNA AK002522.1 - 3 520 145
          AK013725.1 - 3 522 145
          AK082923.1 - 3 520 145
          AK147977.1 - 3 520 145
          BC008106.1 - 3 508 3158148 145
          BC034258.1 - 3 508 3158148 145
          BC085492.1 - 1 2108 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dynll1

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transport
          apoptotic process
          microtubule-based process
          microtubule-based movement
          intraciliary retrograde transport
          negative regulation of phosphorylation
          negative regulation of catalytic activity
          negative regulation of nitric oxide biosynthetic process
          regulation of catalytic activity
          cilium morphogenesis
          motor activity
          enzyme inhibitor activity
          protein binding
          protein C-terminus binding
          enzyme binding
          protein domain specific binding
          nitric-oxide synthase regulator activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008929 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008926 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 AK147977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 BF456225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 AK082923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 NM_019682.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 AK147977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 BF456225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 AK082923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P250555 NM_019682.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47352 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169297 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 AK147977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 BF456225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 AK082923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 NM_019682.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 AK147977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 BF456225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 AK082923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P250555 NM_019682.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414518 BC008106.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414518 BC034258.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414518 BC085492.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414518 AK147977.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414518 AK010614.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008915 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169297 - -
          Whole Mouse Genome G4122A A_51_P414518 BC008106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414518 BC034258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414518 BC085492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414518 AK147977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414518 AK010614.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104002 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04101507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04126837 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04126833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04126840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04126839 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04126838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04218768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04769716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04379952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04597622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04379954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04597620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04597614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04769713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04126830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008919 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532985 - -
          Affymetrix HT MG-430 PM Array Plate 1448682_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440278_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608684 - -
          Affymetrix Mouse Gene 1.1 ST Array 10532984 - -
          Mouse Genome 430 2.0 Array 1448682_PM_at - -
          Mouse Genome 430 2.0 Array 1417339_a_at - -
          Mouse Genome 430 2.0 Array 1417339_PM_a_at - -
          Mouse Genome 430 2.0 Array 1456125_PM_a_at - -
          Mouse Genome 430 2.0 Array 1440278_PM_at - -
          Mouse Genome 430 2.0 Array 1456125_a_at - -
          Mouse Genome 430 2.0 Array 1440278_at - -
          Mouse Genome 430 2.0 Array 1448682_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56455_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56455_at - -
          Affymetrix Mouse Clariom S Array PSR0500027570.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027570.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027570.mm.1 NM_019682.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027571.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027571.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027571.mm.1 NM_019682.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027568.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027568.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027568.mm.1 NM_019682.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027574.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027574.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027574.mm.1 NM_019682.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027561.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027561.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027561.mm.1 NM_019682.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027563.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027563.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027563.mm.1 NM_019682.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027564.mm.1 TC0500003095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027564.mm.1 NM_019682.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027564.mm.1 NM_019682.4 Refseq
          Mouse Expression Set 430 1448682_PM_at - -
          Mouse Expression Set 430 1456125_a_at - -
          Mouse Expression Set 430 1417339_a_at - -
          Mouse Expression Set 430 1417339_PM_a_at - -
          Mouse Expression Set 430 1456125_PM_a_at - -
          Mouse Expression Set 430 1448682_at - -
          Mouse Expression Set 430 1440278_PM_at - -
          Mouse Expression Set 430 1440278_at - -
          Murine Genome U74v2 Set 96298_PM_f_at - -
          Murine Genome U74v2 Set 96298_f_at - -
          Mouse Genome 430A 2.0 Array 1448682_at - -
          Mouse Genome 430A 2.0 Array 1448682_PM_at - -
          Mouse Genome 430A 2.0 Array 1417339_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1456125_a_at - -
          Mouse Genome 430A 2.0 Array 1456125_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417339_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448682_at - -
          Affymetrix HT Mouse Genome 430A Array 1448682_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440278_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440278_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 351403 - -
          ABI Mouse Genome Survey Microarray 894755 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2776422 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2776422 - -
          mouse-6 v1.1 (Illumina) scl056455.3_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056455.3_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2788121 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2788121 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2776419 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056455.3_23-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline