Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850353_g1
          See other OLFR141 GE Assays ›
          Gene Symbol:
          Olfr141
          Gene Name:
          olfactory receptor 141
          Gene Aliases:
          K17, MOR179-5
          Chromosome Location:
          Chr.2: 86806021 - 86806997 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00850353_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr141
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181818.2 NP_861539.2 1 560 143

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr141

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424290 NM_181818.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424290 NM_181818.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424290 NM_181818.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424290 NM_181818.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403673 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139194 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100442 NM_146730.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01431488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01570115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01570112 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01570121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01916486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01570115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02276145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01785235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01431489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01785223 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484633 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484632 - -
          Affymetrix Mouse Clariom S Array PSR0200033809.mm.1 NM_181818.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033809.mm.1 NM_181818.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033809.mm.1 TC0200003957.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033810.mm.1 NM_181818.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033810.mm.1 NM_181818.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033810.mm.1 TC0200003957.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033808.mm.1 NM_181818.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033808.mm.1 NM_181818.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033808.mm.1 TC0200003957.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 420333 - -
          ABI Mouse Genome Survey Microarray 543819 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2350019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2611205 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2611205 - -
          mouse-6 v1.1 (Illumina) scl00257913.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00257913.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2611205 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611205 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline