Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00851399_s1
          See other CHCHD4 GE Assays ›
          Gene Symbol:
          Chchd4
          Gene Name:
          coiled-coil-helix-coiled-coil-helix domain containing 4
          Gene Aliases:
          2410012P20Rik, 2810014D17Rik, AI838740
          Chromosome Location:
          Chr.6: 91464276 - 91473423 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00851399_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chchd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133928.2 NP_598689.1 3 959 112
          GenBank mRNA AK010479.1 - 3 930 112
          BC019405.1 - 3 959 5066665 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chchd4

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          protein maturation by protein folding
          cellular response to oxidative stress
          mitochondrial DNA repair
          protein import into mitochondrial intermembrane space
          regulation of protein export from nucleus
          'de novo' posttranslational protein folding
          oxidation-reduction process
          establishment of protein localization to mitochondrion
          positive regulation of cellular respiration
          protein binding
          protein disulfide oxidoreductase activity
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639203 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254234 BC019405.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254234 NM_133928.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254234 AK010479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254234 BC019405.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254234 NM_133928.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254234 AK010479.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130651 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171277 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254234 BC019405.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254234 NM_133928.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254234 AK010479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254234 BC019405.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254234 NM_133928.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254234 AK010479.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254234 BC019405.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254234 NM_133928.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254234 AK010479.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254234 BC019405.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254234 NM_133928.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254234 AK010479.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639174 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171277 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01384059 - -
          Whole Mouse Genome G4122A A_51_P254234 BC019405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254234 NM_133928.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254234 AK010479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254234 BC019405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254234 NM_133928.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254234 AK010479.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129677 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171277 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639203 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05261494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05142624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05142629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05850278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05594503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05142645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05261491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05726742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05726740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05091243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05623964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05142642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10546348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546347 - -
          Affymetrix HT MG-430 PM Array Plate 1417233_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10546346 - -
          Mouse Genome 430 2.0 Array 1417233_at - -
          Mouse Genome 430 2.0 Array 1417233_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72170_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72170_PM_at - -
          Mouse Expression Set 430 1417233_PM_at - -
          Mouse Expression Set 430 1417233_at - -
          Murine Genome U74v2 Set 168595_r_at - -
          Murine Genome U74v2 Set 168091_PM_f_at - -
          Murine Genome U74v2 Set 168595_PM_r_at - -
          Murine Genome U74v2 Set 168510_PM_f_at - -
          Murine Genome U74v2 Set 168091_f_at - -
          Murine Genome U74v2 Set 168510_f_at - -
          Mouse Genome 430A 2.0 Array 1417233_at - -
          Mouse Genome 430A 2.0 Array 1417233_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417233_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417233_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 343919 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2680075 - -
          Illumina mouse-6 v1.0 expression beadchip 2680075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2624695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2624695 - -
          mouse-6 v1.1 (Illumina) scl28683.3_131-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28683.3_131-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2624695 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2624695 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28683.3_131-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline