Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00918640_gH
          See other CSDE1 GE Assays ›
          Gene Symbol:
          Csde1
          Gene Name:
          cold shock domain containing E1, RNA binding
          Gene Aliases:
          AA960392, BC016898, D3Jfr1, mKIAA0885, unr
          Chromosome Location:
          Chr.3: 103020431 - 103058189 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00918640_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csde1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161854.1 NP_001155326.1 12 - 13 1919 79
          NM_144901.3 NP_659150.1 13 - 14 2012 79
          GenBank mRNA AK122393.1 - 12 - 13 1829 79
          AK145351.1 - 12 - 13 2024 79
          AK145428.1 - 12 - 13 1841 79
          AK145932.1 - 12 - 13 1836 79
          AK146586.1 - 12 - 13 1860 79
          AK146706.1 - 13 - 14 1944 79
          AK147012.1 - 12 - 13 1858 79
          AK150882.1 - 12 - 13 1837 79
          AK151638.1 - 12 - 13 1855 79
          AK151723.1 - 12 - 13 1859 79
          AK151970.1 - 12 - 13 1859 79
          AK152531.1 - 12 - 13 1855 79
          AK152777.1 - 8 - 9 1066 79
          AK153196.1 - 12 - 13 1836 79
          AK153263.1 - 8 - 9 1066 79
          AK154068.1 - 12 - 13 1844 79
          AK155751.1 - 13 - 14 1937 79
          AK159258.1 - 12 - 13 1852 79
          AK159863.1 - 12 - 13 1852 79
          AK168556.1 - 11 - 12 1655 79
          BC016898.1 - 12 - 13 1856 4224466 79
          BC024826.1 - 9 - 10 1139 79
          BC029058.1 - 7 - 8 798 4947555 79
          BC029688.1 - 12 - 13 1855 5367783 79
          BC031404.1 - 12 - 13 1827 4953772 79
          BC036720.1 - 9 - 10 1145 79
          BC055072.1 - 2855 79
          BC062097.1 - 13 - 14 1927 6833505 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001040.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csde1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nuclear-transcribed mRNA catabolic process, no-go decay
          nucleic acid binding
          DNA binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300008341.mm.1 NM_001161854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008341.mm.1 NM_144901.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008341.mm.1 TC0300001040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008342.mm.1 NM_001161854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008342.mm.1 NM_144901.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008342.mm.1 TC0300001040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008343.mm.1 NM_001161854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008343.mm.1 NM_144901.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008343.mm.1 TC0300001040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008338.mm.1 NM_001161854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008338.mm.1 NM_144901.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008338.mm.1 TC0300001040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008340.mm.1 NM_001161854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008340.mm.1 NM_144901.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008340.mm.1 TC0300001040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300008339.mm.1 NM_001161854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008339.mm.1 NM_144901.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300008339.mm.1 TC0300001040.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline