Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01132688_m1
          See other PTN GE Assays ›
          Gene Symbol:
          Ptn
          Gene Name:
          pleiotrophin
          Gene Aliases:
          HARP, HB-GAM, HBBN, HBGF-8, HBNF, OSF, Osf-1, Osf1
          Chromosome Location:
          Chr.6: 36714909 - 36811361 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01132688_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008973.2 NP_032999.1 3 - 4 1758 61
          GenBank mRNA AK011346.1 - 3 - 4 615 61
          AK011699.1 - 3 - 4 541 61
          AK017768.1 - 3 - 4 577 61
          AK133835.1 - 3 - 4 558 61
          AK161847.1 - 3 - 4 508 61
          AK161964.1 - 3 - 4 448 61
          BC002064.1 - 3 - 4 327 3488877 61
          BC048363.1 - 3 - 4 332 61
          BC061695.1 - 3 - 4 558 6313227 61
          D90225.1 - 3 - 4 502 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002079.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptn

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          neuromuscular junction development
          learning
          positive regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          bone mineralization
          negative regulation of membrane potential
          positive regulation of cell division
          retinal rod cell differentiation
          skeletal muscle acetylcholine-gated channel clustering
          positive regulation of hepatocyte proliferation
          glycosaminoglycan binding
          growth factor activity
          heparin binding
          chondroitin sulfate proteoglycan binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600016343.mm.1 TC0600002079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016343.mm.1 NM_008973.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016343.mm.1 NM_008973.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016348.mm.1 TC0600002079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016348.mm.1 NM_008973.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016348.mm.1 NM_008973.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016347.mm.1 TC0600002079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016347.mm.1 NM_008973.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016347.mm.1 NM_008973.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016345.mm.1 TC0600002079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016345.mm.1 NM_008973.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016345.mm.1 NM_008973.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016346.mm.1 TC0600002079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016346.mm.1 NM_008973.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016346.mm.1 NM_008973.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016344.mm.1 TC0600002079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016344.mm.1 NM_008973.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016344.mm.1 NM_008973.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 907116 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline