Search
Search

Camk4| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009793.3 | NP_033923.2 | 4 - 5 | 730 | 65 | |
| GenBank mRNA | AK030219.1 | - | 4 - 5 | 614 | 65 | |
| AK043114.1 | - | 4 - 5 | 440 | 65 | ||
| BC070420.1 | - | 4 - 5 | 611 | 5718423 | 65 | |
| X58995.1 | - | 4 - 5 | 420 | 65 | ||
Set Membership: |
|