Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01136095_m1
          See other HFE GE Assays ›
          Gene Symbol:
          Hfe
          Gene Name:
          hemochromatosis
          Gene Aliases:
          MR2
          Chromosome Location:
          Chr.13: 23703564 - 23710986 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01136095_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hfe
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010424.4 NP_034554.2 2 - 3 450 80
          GenBank mRNA AK009581.1 - 2 - 3 451 80
          AK088986.1 - 2 - 3 446 80
          AK150697.1 - 2 - 3 468 80
          BC116744.1 - 2 - 3 408 40126063 80
          BC116746.1 - 2 - 3 406 40126065 80
          U66849.1 - 2 - 3 432 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001641.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hfe

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          antigen processing and presentation of peptide antigen via MHC class I
          immune response
          positive regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          antigen processing and presentation
          BMP signaling pathway
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          hormone biosynthetic process
          iron ion homeostasis
          multicellular organismal iron ion homeostasis
          iron ion import into cell
          negative regulation of receptor binding
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          antigen binding
          receptor binding
          co-receptor binding
          peptide antigen binding
          transferrin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056070 AK150697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056070 AJ306425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056070 AK088986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056070 NM_010424.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056070 AJ298838.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568193 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121726 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056070 AK150697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056070 AJ306425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056070 AK088986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056070 NM_010424.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056070 AJ298838.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P193022 AJ306425.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505538 AK150697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505538 U66849.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505538 AJ306425.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505538 AK009581.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505538 AK088986.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883042 - -
          Whole Mouse Genome G4122A A_52_P193022 AJ306425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505538 AK150697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505538 U66849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505538 AJ306425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505538 AK009581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505538 AK088986.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136514 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10627186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11012948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11012950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11012952 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10508079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10513342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10639905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10676906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10481393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11010754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11060716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10676909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10530252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10627185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11012952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11047384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11047383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408228 - -
          Affymetrix HT MG-430 PM Array Plate 1450702_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422645_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408227 - -
          Mouse Genome 430 2.0 Array 1422645_PM_at - -
          Mouse Genome 430 2.0 Array 1450702_PM_at - -
          Mouse Genome 430 2.0 Array 1450702_at - -
          Mouse Genome 430 2.0 Array 1422645_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15216_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15216_at - -
          Affymetrix Mouse Clariom S Array PSR1300010175.mm.1 NM_010424.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010175.mm.1 TC1300001641.mm.1 MTA TC
          Mouse Expression Set 430 1450702_at - -
          Mouse Expression Set 430 1422645_PM_at - -
          Mouse Expression Set 430 1422645_at - -
          Mouse Expression Set 430 1450702_PM_at - -
          Murine Genome U74v2 Set 104014_PM_at - -
          Murine Genome U74v2 Set 104014_at - -
          Mouse Genome 430A 2.0 Array 1422645_PM_at - -
          Mouse Genome 430A 2.0 Array 1422645_at - -
          Mouse Genome 430A 2.0 Array 1450702_at - -
          Mouse Genome 430A 2.0 Array 1450702_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422645_at - -
          Affymetrix HT Mouse Genome 430A Array 1422645_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450702_at - -
          Affymetrix HT Mouse Genome 430A Array 1450702_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 341817 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5550059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2592519 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2592519 - -
          mouse-6 v1.1 (Illumina) scl44206.4_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44206.4_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2870696 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2592519 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2870696 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2592519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0