| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
TC0600000289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040257.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040257.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040260.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040260.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040261.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040261.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040262.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002097.mm.1 |
NR_040262.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
TC0600000289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040257.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040257.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040260.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040260.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040261.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040261.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040262.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002098.mm.1 |
NR_040262.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
TC0600000289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040257.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040257.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040261.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040261.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040262.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600002105.mm.1 |
NR_040262.1 |
Refseq |