Search
Search

Glcci1| Gene Symbol: Glcci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286728.1 | NP_001273657.1 | 6 - 7 | 768 | 92 | |
| NM_001286729.1 | NP_001273658.1 | 6 - 7 | 765 | 92 | ||
| NM_133236.3 | NP_573499.2 | 6 - 7 | 1441 | 92 | ||
| GenBank mRNA | AK029524.1 | - | 6 - 7 | 767 | 92 | |
| AY098636.1 | - | 6 - 7 | 1487 | 92 | ||
| BC050802.1 | - | 6 - 7 | 806 | 6771009 | 92 | |
| DV652660.1 | - | 2 - 3 | 340 | 92 | ||
Set Membership: |
|