Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01137654_g1
          See other NDUFAB1 GE Assays ›
          Gene Symbol:
          Ndufab1
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1
          Gene Aliases:
          2210401F17Rik, 2310039H15Rik, 2610003B19Rik, 8kDa, 9130423F15Rik, ACP, CI-SDAP, SDAP
          Chromosome Location:
          Chr.7: 122088044 - 122101877 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01137654_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufab1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028177.3 NP_082453.2 1 - 2 195 97
          GenBank mRNA AK008788.1 - 192 97
          AK009698.1 - 1 - 2 197 97
          AK010307.1 - 1 - 2 196 97
          AK011302.1 - 1 - 2 196 97
          AK018688.1 - 1 - 2 195 97
          AK018717.1 - 1 - 2 191 97
          AK089103.1 - 1 - 2 185 97
          AK151719.1 - 1 - 2 195 97
          AK152995.1 - 1 - 2 195 97
          AK167447.1 - 1 - 2 176 97
          BC060951.1 - 1 - 2 188 6774795 97
          BC092379.1 - 1 - 2 184 6815043 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufab1

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          transport
          protein lipoylation
          oxidation-reduction process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700035174.mm.1 NM_028177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035174.mm.1 NM_028177.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035174.mm.1 TC0700004224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035178.mm.1 NM_028177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035178.mm.1 NM_028177.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035178.mm.1 TC0700004224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035167.mm.1 TC0700004224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035170.mm.1 NM_028177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035170.mm.1 NM_028177.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035170.mm.1 TC0700004224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035181.mm.1 NM_028177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035181.mm.1 NM_028177.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035181.mm.1 TC0700004224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035183.mm.1 NM_028177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035183.mm.1 NM_028177.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035183.mm.1 TC0700004224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035180.mm.1 NM_028177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035180.mm.1 NM_028177.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035180.mm.1 TC0700004224.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895303 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline