| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700014411.mm.1 |
NM_028129.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014411.mm.1 |
NM_028129.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014411.mm.1 |
TC0700001716.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014419.mm.1 |
NM_001004187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014419.mm.1 |
NM_001004187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014419.mm.1 |
NM_028129.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014419.mm.1 |
NM_028129.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014419.mm.1 |
TC0700001716.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014420.mm.1 |
NM_001004187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014420.mm.1 |
NM_001004187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014420.mm.1 |
NM_028129.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014420.mm.1 |
NM_028129.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014420.mm.1 |
TC0700001716.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014421.mm.1 |
NM_001004187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014421.mm.1 |
NM_001004187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014421.mm.1 |
NM_028129.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014421.mm.1 |
NM_028129.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014421.mm.1 |
TC0700001716.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014418.mm.1 |
NM_001004187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014418.mm.1 |
NM_001004187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014418.mm.1 |
NM_028129.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014418.mm.1 |
NM_028129.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014418.mm.1 |
TC0700001716.mm.1 |
MTA TC |