Search
Search

Pmpcb| Gene Symbol: Pmpcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028431.2 | NP_082707.1 | 8 - 9 | 1005 | 90 | |
| GenBank mRNA | AK013995.1 | - | 8 - 9 | 1006 | 90 | |
| AK152673.1 | - | 8 - 9 | 1006 | 90 | ||
| AK169417.1 | - | 8 - 9 | 962 | 90 | ||
| BC038321.1 | - | 7 - 8 | 876 | 90 | ||
| BC051216.1 | - | 7 - 8 | 812 | 90 | ||
| BC060662.1 | - | 7 - 8 | 3517 | 90 | ||
| BC065095.1 | - | 7 - 8 | 3517 | 90 | ||
| BC087943.1 | - | 8 - 9 | 1017 | 90 | ||
Set Membership: |
|