Search
Search

Kctd13| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172747.2 | NP_766335.1 | 1 - 2 | 423 | 90 | |
| GenBank mRNA | AF534881.3 | - | 1 - 2 | 409 | 90 | |
| AK075722.1 | - | 1 - 2 | 424 | 90 | ||
| BC079607.1 | - | 1 - 2 | 416 | 30359035 | 90 | |
Set Membership: |
|