Search
Search

Tmem123| Gene Symbol: Tmem123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133739.2 | NP_598500.1 | 1 - 2 | 234 | 78 | |
| GenBank mRNA | AK150361.1 | - | 1 - 2 | 182 | 78 | |
| AK150365.1 | - | 1 - 2 | 146 | 78 | ||
| AK151044.1 | - | 1 - 2 | 202 | 78 | ||
| AK152180.1 | - | 1 - 2 | 182 | 78 | ||
| AK152423.1 | - | 1 - 2 | 147 | 78 | ||
| AK153122.1 | - | 1 - 2 | 147 | 78 | ||
| AK153378.1 | - | 1 - 2 | 146 | 78 | ||
| AK153539.1 | - | 1 - 2 | 182 | 78 | ||
| AK155767.1 | - | 1 - 2 | 202 | 78 | ||
| AK162610.1 | - | 1 - 2 | 230 | 78 | ||
| AK165671.1 | - | 1 - 2 | 201 | 78 | ||
| AK170823.1 | - | 1 - 2 | 193 | 78 | ||
| AK172352.1 | - | 1 - 2 | 163 | 78 | ||
| AK172553.1 | - | 1 - 2 | 163 | 78 | ||
| AK172701.1 | - | 1 - 2 | 163 | 78 | ||
| BC010292.1 | - | 1 - 2 | 183 | 3153174 | 78 | |
| BC086319.1 | - | 1 - 2 | 191 | 5293409 | 78 | |
Set Membership: |
|