Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01143609_m1
          See other NLRP5 GE Assays ›
          Gene Symbol:
          Nlrp5
          Gene Name:
          NLR family, pyrin domain containing 5
          Gene Aliases:
          Mater, Nalp5, Op1, PAN11
          Chromosome Location:
          Chr.7: 23385897 - 23441921 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01143609_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlrp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF074018.1 - 14 - 15 3211 71
          AK087774.1 - 14 - 15 3233 71
          AY196361.1 - 14 - 15 3211 71
          AY196362.1 - 14 - 15 3211 71
          AY329484.1 - 13 - 14 3163 71
          AY329485.1 - 14 - 15 3129 71
          AY329486.1 - 14 - 15 3211 71
          AY329487.1 - 13 - 14 3163 71
          AY329488.1 - 13 - 14 3061 71
          AY329489.1 - 14 - 15 3133 71
          AY329490.1 - 14 - 15 3211 71
          AY329491.1 - 13 - 14 3061 71
          BC053384.1 - 15 - 16 3278 30049556 71
          BG071729.2 - 1 - 2 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000351.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987151 AY329484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987151 AY196361.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356445 BC003705.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356445 L40156.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356445 NM_009160.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155841 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101190 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152249 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112977 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987151 AY329484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987151 AY196361.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415293 BC053384.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415293 AY329490.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415293 AY329485.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415293 AY329486.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415293 AY329484.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25001551 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03806010 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152249 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03806024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06139210 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06139202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112977 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06139240 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101190 - -
          Whole Mouse Genome G4122A A_51_P415293 BC053384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415293 AY329490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415293 AY329485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415293 AY329486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415293 AY329484.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117412 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06139202 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152249 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112977 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03806024 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25001527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25001568 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550799 - -
          Affymetrix HT MG-430 PM Array Plate 1418882_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550795 - -
          Mouse Genome 430 2.0 Array 1418882_PM_at - -
          Mouse Genome 430 2.0 Array 1418882_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23968_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23968_at - -
          Affymetrix Mouse Clariom S Array PSR0700002612.mm.1 TC0700000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002612.mm.1 NM_001039143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002612.mm.1 NM_011860.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002610.mm.1 TC0700000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002610.mm.1 NM_001039143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002610.mm.1 NM_011860.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002609.mm.1 TC0700000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002609.mm.1 NM_001039143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002609.mm.1 NM_011860.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002611.mm.1 TC0700000351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002611.mm.1 NM_001039143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002611.mm.1 NM_011860.3 Refseq
          Mouse Expression Set 430 1418882_PM_at - -
          Mouse Expression Set 430 1418882_at - -
          Mouse Genome 430A 2.0 Array 1418882_at - -
          Mouse Genome 430A 2.0 Array 1418882_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418882_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418882_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170619 - -
          Illumina mouse-6 v1.0 expression beadchip 6350131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646011 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696556 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696556 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646011 - -
          mouse-6 v1.1 (Illumina) scl32977.8_62-S - -
          mouse-6 v1.1 (Illumina) scl023968.15_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350131 - -
          Illumina Sentrix mouse-6.v1 bead array 3170619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32977.8_62-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl023968.15_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350131 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646011 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696556 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3152906 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3152906 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646011 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696556 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline