Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01144210_m1
          See other NDUFS7 GE Assays ›
          Gene Symbol:
          Ndufs7
          Gene Name:
          NADH dehydrogenase (ubiquinone) Fe-S protein 7
          Gene Aliases:
          1010001M04Rik
          Chromosome Location:
          Chr.10: 80249121 - 80256793 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01144210_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufs7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029272.3 NP_083548.1 6 - 7 515 69
          GenBank mRNA AK003132.1 - 6 - 7 516 69
          BC013503.1 - 6 - 7 492 4223916 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000847.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufs7

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          mitochondrial respiratory chain complex I assembly
          oxidation-reduction process
          protease binding
          NADH dehydrogenase activity
          NADH dehydrogenase (ubiquinone) activity
          oxidoreductase activity
          metal ion binding
          quinone binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281951 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281950 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281948 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281949 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974273 BC013503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974273 NM_029272.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24932 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140203 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974273 BC013503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974273 NM_029272.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364671 AK003132.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27281947 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02168660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140203 - -
          Whole Mouse Genome G4122A A_51_P364671 AK003132.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114284 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140203 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08530793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08501907 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08501908 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08501917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08753741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281951 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281948 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281950 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27281949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08649533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08530793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08501909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08517980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08519952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08261379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08649544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08596592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08847520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08501912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08261374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08519956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27281941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08649545 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364749 - -
          Affymetrix HT MG-430 PM Array Plate 1424313_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1451312_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364744 - -
          Mouse Genome 430 2.0 Array 1451312_at - -
          Mouse Genome 430 2.0 Array 1424313_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451312_PM_at - -
          Mouse Genome 430 2.0 Array 1424313_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75406_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75406_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000006421.mm.1 NM_029272.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006421.mm.1 NM_029272.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006421.mm.1 TC1000000847.mm.1 MTA TC
          Mouse Expression Set 430 1451312_at - -
          Mouse Expression Set 430 1424313_PM_a_at - -
          Mouse Expression Set 430 1424313_a_at - -
          Mouse Expression Set 430 1451312_PM_at - -
          Mouse Genome 430A 2.0 Array 1451312_PM_at - -
          Mouse Genome 430A 2.0 Array 1451312_at - -
          Mouse Genome 430A 2.0 Array 1424313_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424313_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451312_at - -
          Affymetrix HT Mouse Genome 430A Array 1424313_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451312_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424313_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 628243 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1740142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688763 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688763 - -
          mouse-6 v1.1 (Illumina) scl38688.8.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38688.8.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2688763 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2688763 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38688.8.1_12-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline