Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01144370_m1
          See other ZFP949 GE Assays ›
          Gene Symbol:
          Zfp949
          Gene Name:
          zinc finger protein 949
          Gene Aliases:
          4930422I07Rik
          Chromosome Location:
          Chr.9: 88548020 - 88571086 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01144370_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp949
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142943.1 NP_001136415.1 5 - 6 1022 72
          NM_001163061.1 NP_001156533.1 4 - 5 876 72
          GenBank mRNA AK015179.1 - 5 - 6 1024 72
          BC141009.1 - 5 - 6 859 9055621 72
          BC141010.1 - 4 - 5 713 9055622 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001132.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513244 AK015179.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513244 AK015179.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170536 BC141009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170536 BC141010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170536 AK137745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170536 NM_001142943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170536 NM_001163061.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101840 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158736 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176051 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156479 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170536 BC141009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170536 BC141010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170536 AK137745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170536 NM_001142943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170536 NM_001163061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513244 AK015179.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513244 AK015179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513244 AK015179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513244 AK015179.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07855837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07924352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08115052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07974663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07706981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07706978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08067633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07706977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07855835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07637323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08067626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26710634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10587656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587658 - -
          Affymetrix HT MG-430 PM Array Plate 1458491_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456518_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10587655 - -
          Mouse Genome 430 2.0 Array 1445654_PM_at - -
          Mouse Genome 430 2.0 Array 1445654_at - -
          Mouse Genome 430 2.0 Array 1458491_PM_at - -
          Mouse Genome 430 2.0 Array 1458491_at - -
          Mouse Genome 430 2.0 Array 1456518_PM_at - -
          Mouse Genome 430 2.0 Array 1456518_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71640_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71640_at - -
          Affymetrix Mouse Clariom S Array PSR0900008888.mm.1 NM_001142943.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008888.mm.1 NM_001142943.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008888.mm.1 NM_001163061.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008888.mm.1 NM_001163061.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008888.mm.1 TC0900001132.mm.1 MTA TC
          Mouse Expression Set 430 1456518_at - -
          Mouse Expression Set 430 1458491_at - -
          Mouse Expression Set 430 1458491_PM_at - -
          Mouse Expression Set 430 1456518_PM_at - -
          Murine Genome U74v2 Set 166784_at - -
          Murine Genome U74v2 Set 166784_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458491_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456518_at - -
          Affymetrix HT Mouse Genome 430B Array 1458491_at - -
          Affymetrix HT Mouse Genome 430B Array 1456518_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228464 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228464 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline