Search
Search

Cldnd1| Gene Symbol: Cldnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252450.1 | NP_001239379.1 | 3 - 4 | 532 | 100 | |
| NM_001252451.1 | NP_001239380.1 | 3 - 4 | 532 | 100 | ||
| NM_171826.3 | NP_741968.1 | 3 - 4 | 539 | 100 | ||
| NR_045517.1 | 2 - 3 | 488 | 100 | |||
| NR_045518.1 | 2 - 3 | 488 | 100 | |||
| GenBank mRNA | AK003808.1 | - | 2 - 3 | 380 | 100 | |
| AK009432.1 | - | 2 - 3 | 352 | 100 | ||
| AK009743.1 | - | 2 - 3 | 372 | 100 | ||
| AK152121.1 | - | 2 - 3 | 383 | 100 | ||
| AK152722.1 | - | 2 - 3 | 378 | 100 | ||
| BC055695.1 | - | 1 - 2 | 296 | 6414170 | 100 | |
| CA750661.1 | - | 3 - 4 | 382 | 100 | ||
| CD560460.1 | - | 2 - 3 | 369 | 100 | ||
Set Membership: |
|