Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam81a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029784.2 | NP_084060.1 | 6 - 7 | 968 | 96 | |
GenBank mRNA | AK038532.1 | - | 6 - 7 | 927 | 96 | |
AK044090.1 | - | 6 - 7 | 968 | 96 | ||
AK046408.1 | - | 6 - 7 | 861 | 96 | ||
AK079541.1 | - | 6 - 7 | 928 | 96 | ||
AK135091.1 | - | 5 - 6 | 683 | 96 | ||
BC025646.1 | - | 6 - 7 | 994 | 5324986 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |