Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008793.2 | NP_032819.1 | 11 - 12 | 1405 | 95 | |
GenBank mRNA | AK161504.1 | - | 9 - 10 | 1664 | 95 | |
D01093.1 | - | 11 - 12 | 1405 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Detect Genomic DNA Mus musculus Probe spans exons |